1 - 25 of 25 products displayed
  • KIF2A Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mammal, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA316833
  • Size 100 µL
Applications
Immunocytochemistry (1:10,000), Immunofluorescence (1:10,000), Immunoprecipitation (1:10-1:500), Western Blot (1:10,000)
Alternate names:
Kinesin-related protein xKIF2; Kinesin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA316833 349.00
Check your price
  • KIF2A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA540802
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.2-1 µg/ml)
Alternate names:
Kinesin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540802 309.00
Check your price
  • KIF5C Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # 408100
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Kinesin heavy chain neuron-specific 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
408100 309.00
Check your price
  • KIF2A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA1640
  • Size 200 µL
Applications
Western Blot (1:200)
Alternate names:
Kinesin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1640 349.00
Check your price
  • KIF2A Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA132454
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (1:10,000), Immunoprecipitation (1:10,000), Western Blot (1:10,000)
Alternate names:
Kinesin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132454 425.00
Check your price
  • KIF5B Polyclonal Antibody for Western Blot and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Pig
  • Conjugate Unconjugated
  • Catalog # PA119392
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA119392 275.00
Check your price
  • KIF5A Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528143
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Neuronal kinesin heavy chain; Kinesin heavy chain neuron-specific 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528143 349.00
Check your price
  • KIF5A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529820
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Neuronal kinesin heavy chain; Kinesin heavy chain neuron-specific 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529820 349.00
Check your price
  • KIF5B Monoclonal Antibody for ICC
  • Host Mouse
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA119353
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119353 225.00
Check your price
  • KIF5B Monoclonal Antibody for IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA119352
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (1:100-1:500)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119352 275.00
Check your price
  • KIF5A Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Bovine, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1642
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Neuronal kinesin heavy chain; Kinesin heavy chain neuron-specific 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1642 375.00
Check your price
  • KIF5B Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1643
  • Size 100 µg
Applications
Immunocytochemistry (1:250), Immunofluorescence (1:250), Immunoprecipitation (5 µg), Western Blot (1:4000-1:6000 )
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1643 375.00
Check your price
  • KIF5C Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1644
  • Size 100 µg
Applications
Western Blot (0.5 ug/ml)
Alternate names:
Kinesin heavy chain neuron-specific 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1644 375.00
Check your price
  • KIF5B Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Dog, Human
  • Conjugate Unconjugated
  • Catalog # PA518063
  • Size 100 ug
Applications
Immunohistochemistry (1-2µg/ml), Western Blot (0.2-1 ug/ml)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518063 349.00
Check your price
  • KIF5A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520420
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Neuronal kinesin heavy chain; Kinesin heavy chain neuron-specific 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520420 349.00
Check your price
  • KIF5A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520421
  • Size 100 ug
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (20 ug/ml), Western Blot (0.5-1 ug/ml)
Alternate names:
Neuronal kinesin heavy chain; Kinesin heavy chain neuron-specific 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520421 349.00
Check your price
  • KIF5B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131243
  • Size 100 µg
Applications
ELISA (1:32,000), Immunohistochemistry (Paraffin) (1-3 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131243 399.00
Check your price
  • KIF5B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521623
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Ubiquitous kinesin heavy chain; Conventional kinesin heavy chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521623 349.00
Check your price
  • KIF1A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540801
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Unc-104- and KIF1A-related protein; Microtubule-based motor KIF1A; Axonal transporter of synaptic vesicles
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540801 309.00
Check your price
  • KIF4A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA131786
  • Size 100 µg
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Western Blot (1:100)
Alternate names:
Chromokinesin; Chromokinesin-A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131786 449.00
Check your price
  • KLC3 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA525268
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
kinesin light chain 2; KLC2-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525268 349.00
Check your price
  • DNAI1 Monoclonal Antibody for Western Blot, IF, ICC, IP and ELISA
  • Host Mouse
  • Target Species Chicken, Deer, Hamster, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA122146
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:2000)
Alternate names:
Axonemal dynein intermediate chain 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA122146 399.00
Check your price
  • NEFH Monoclonal Antibody for Western Blot, IHC (F) and ELISA
  • Host Mouse
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA110441
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunohistochemistry (Frozen) (Assay-Dependent), Western Blot (1 ug/ml)
Alternate names:
Neurofilament triplet H protein; 200 kDa neurofilament protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110441 225.00
Check your price
  • alpha Synuclein Oligoclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710110
  • Size 100 µg
Applications
Immunocytochemistry (1:500-1:5000), Immunofluorescence (1:500-1:5000), Western Blot (1:500-1:5000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
710110 309.00
Check your price
  • alpha Synuclein Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701085
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:500), Immunocytochemistry (1:500-1:5000), Immunofluorescence (1:500-1:5000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:5000)
Alternate names:
Non-A4 component of amyloid precursor; Non-A beta component of AD amyloid
Catalog # Size Price (AUD) Your Price (AUD) Qty
701085 304.00
Check your price