1 - 3 of 3 products displayed
  • KLF1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528132
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Erythroid krueppel-like transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528132 349.00
Check your price
  • KLF1 Polyclonal Antibody for ChIP
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518031
  • Size 100µg
Applications
ChIP assay (Assay Dependent)
Alternate names:
Erythroid krueppel-like transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518031 349.00
Check your price
  • KLF1 Oligoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710917
  • Size 100µg
Applications
Flow Cytometry (2-4 µg/million cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
Erythroid krueppel-like transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710917 309.00
Check your price