1 - 3 of 3 products displayed
  • KLF3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530621
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
TEF-2; CACCC-box-binding protein BKLF; Basic krueppel-like factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530621 349.00
Check your price
  • KLF3 Polyclonal Antibody for Western Blot and ELISA
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA131313
  • Size 100µg
Applications
ELISA (Assay-dependent), Western Blot (1-3 µg/ml)
Alternate names:
TEF-2; CACCC-box-binding protein BKLF; Basic krueppel-like factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131313 399.00
Check your price
  • KLF3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518030
  • Size 100ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
TEF-2; CACCC-box-binding protein BKLF; Basic krueppel-like factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518030 349.00
Check your price