1 - 3 of 3 products displayed
  • KLHL13 Monoclonal Antibody for Western Blot, IF, ICC, IHC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA515658
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
BTB and kelch domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515658 349.00
Check your price
  • KLHL13 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA531658
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
BTB and kelch domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531658 349.00
Check your price
  • KLHL13 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534881
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
BTB and kelch domain-containing protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534881 349.00
Check your price