1 - 2 of 2 products displayed
  • KLHL21 Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515654
  • Size 100µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
KIAA0469; MGC99635; KLHL21
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515654 349.00
Check your price
  • KLHL21 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531730
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
kelch-like protein 21
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531730 349.00
Check your price