1 - 5 of 5 products displayed
  • Kallikrein 11 (Cleaved Ile54) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537436
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Trypsin-like protease; Serine protease 20; Hippostasin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537436 309.00
Check your price
  • Kallikrein 11 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547164
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Trypsin-like protease; Serine protease 20; Hippostasin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547164 349.00
Check your price
  • Kallikrein 11 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA528008
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Trypsin-like protease; Serine protease 20; Hippostasin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528008 349.00
Check your price
  • Kallikrein 11 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA14312
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Trypsin-like protease; Serine protease 20; Hippostasin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14312 309.00
Check your price
  • Kallikrein 11 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA146127
  • Size 100µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Trypsin-like protease; Serine protease 20; Hippostasin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146127 309.00
Check your price