1 - 7 of 7 products displayed
  • Kallikrein 5 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518675
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Stratum corneum tryptic enzyme; Kallikrein-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518675 349.00
Check your price
  • Kallikrein 5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513345
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Stratum corneum tryptic enzyme; Kallikrein-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513345 349.00
Check your price
  • Kallikrein 5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA14238
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Stratum corneum tryptic enzyme; Kallikrein-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14238 309.00
Check your price
  • Kallikrein 5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA121722
  • Size 100µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Stratum corneum tryptic enzyme; Kallikrein-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121722 349.00
Check your price
  • Kallikrein 11 (Cleaved Ile54) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537436
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Trypsin-like protease; Serine protease 20; Hippostasin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537436 309.00
Check your price
  • Kallikrein 11 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547164
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Trypsin-like protease; Serine protease 20; Hippostasin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547164 349.00
Check your price
  • SPINK9 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA545975
  • Size 100µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Lymphoepithelial Kazal-type-related inhibitor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545975 309.00
Check your price