1 - 10 of 10 products displayed
  • Klotho Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA129899
  • Size 25 µg
Applications
ELISA (1:10,000-1:100,000), Western Blot (1-10 µg/ml)
Alternate names:
alpha-kl
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129899 399.00
Check your price
  • Klotho Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542021
  • Size 100 µl
Applications
Immunohistochemistry (5 µg/ml), Western Blot (0.2-1 µg/ml)
Alternate names:
Klotho
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542021 309.00
Check your price
  • Klotho Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542022
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Klotho
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542022 309.00
Check your price
  • Klotho Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518447
  • Size 100 ug
Applications
Immunohistochemistry (4-6µg/ml)
Alternate names:
KL; klotho
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518447 349.00
Check your price
  • LCTL Polyclonal Antibody for Western Blot and IP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547801
  • Size 100 µg
Applications
Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Klotho/lactase-phlorizin hydrolase-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547801 349.00
Check your price
  • KLB Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA544023
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (0.5 µg/ml )
Alternate names:
Klotho beta-like protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544023 309.00
Check your price
  • GBA3 Monoclonal Antibody for IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524342
  • Size 100 µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
Cytosolic beta-glucosidase-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524342 275.00
Check your price
  • GBA3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530782
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Cytosolic beta-glucosidase-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530782 349.00
Check your price
  • GBA3 Polyclonal Antibody for Western Blot and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA548095
  • Size 100 µg
Applications
Immunoprecipitation (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Cytosolic beta-glucosidase-like protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA548095 309.00
Check your price
  • FGF23 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA546997
  • Size 100 µg
Applications
Neutralization (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Tumor-derived hypophosphatemia-inducing factor; Phosphatonin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546997 375.00
Check your price