1 - 5 of 5 products displayed
  • KPNA4 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521033
  • Size 100 ug
Applications
Immunocytochemistry (2.5 ug/ml), Immunofluorescence (20 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521033 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511810
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511810 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521749
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521749 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA518239
  • Size 100 ug
Applications
Immunohistochemistry (3µg/ml), Western Blot (0.2-1 ug/ml)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518239 349.00
Check your price
  • KPNA4 Polyclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Goat
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132097
  • Size 100 µg
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (0.2-1 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Karyopherin subunit alpha-4; Importin alpha Q1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132097 349.00
Check your price