1 - 3 of 3 products displayed
  • ZNF23 Antibody (Monoclonal, 2D3)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA520249
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
Zinc finger protein KOX16; Zinc finger protein 612; Zinc finger protein 359
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA520249 309.00
Check your price
  • ZNF23 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA536353
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Zinc finger protein KOX16; Zinc finger protein 612; Zinc finger protein 359
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536353 309.00
Check your price
  • ZNF23 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA538581
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Zinc finger protein KOX16; Zinc finger protein 612; Zinc finger protein 359
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538581 309.00
Check your price