1 - 4 of 4 products displayed
  • KCNC1 Polyclonal Antibody for Western Blot, IP and Neu
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA523012
  • Size 100 µL
Applications
Blocking Assay (50-200 molar excess), Immunoprecipitation (1:10-1:500), Neutralization (50-200 molar excess), Western Blot (1:500-1:1000)
Alternate names:
Voltage-gated potassium channel subunit Kv4; Voltage-gated potassium channel subunit Kv3.1; NGK2; RAW2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523012 349.00
Check your price
  • KCNC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA112659
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Voltage-gated potassium channel subunit Kv4; Voltage-gated potassium channel subunit Kv3.1; NGK2; RAW2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112659 309.00
Check your price
  • KCNC1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542894
  • Size 100 µl
Applications
Western Blot (2.5 µg/ml)
Alternate names:
Voltage-gated potassium channel subunit Kv4; Voltage-gated potassium channel subunit Kv3.1; NGK2; RAW2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542894 309.00
Check your price
  • Phospho-KCNC1 (Ser503) Polyclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA14638
  • Size 100 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (1:1000), Western Blot (1:1000)
Alternate names:
Voltage-gated potassium channel subunit Kv4; Voltage-gated potassium channel subunit Kv3.1; NGK2; RAW2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14638 309.00
Check your price