1 - 23 of 23 products displayed
  • LDHA Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527406
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Renal carcinoma antigen NY-REN-59; LDH muscle subunit; Cell proliferation-inducing gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527406 349.00
Check your price
  • LDHC Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530079
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
LDH-X; LDH testis subunit; Cancer/testis antigen 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530079 349.00
Check your price
  • LDHA Polyclonal Antibody for Western Blot, IF, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA519754
  • Size 100µg
Applications
Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1 µg/ml), Immunoprecipitation (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Renal carcinoma antigen NY-REN-59; LDH muscle subunit; Cell proliferation-inducing gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519754 349.00
Check your price
  • LDHA Polyclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IP and ELISA
  • Host Goat
  • Target Species Rabbit
  • Conjugate Biotin
  • Catalog # PA126911
  • Size 1mg
Applications
ELISA (1:5000-1:20,000), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (1:100), Western Blot (1:500-1:5000)
Alternate names:
LDH muscle subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126911 309.00
Check your price
  • LDHAL6A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524372
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
LDH6A, L-lactate dehydrogenase A-like 6A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524372 349.00
Check your price
  • LDHA Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523036
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1000-1:2500)
Alternate names:
Renal carcinoma antigen NY-REN-59; LDH muscle subunit; Cell proliferation-inducing gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523036 349.00
Check your price
  • LDHA Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526531
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Renal carcinoma antigen NY-REN-59; LDH muscle subunit; Cell proliferation-inducing gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526531 349.00
Check your price
  • LDHD Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA525637
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
DLD, D-lactate dehydrogenase, probable D-lactate dehydrogenase, mitochondrial
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA525637 349.00
Check your price
  • LDHA Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517246
  • Size 100µL
Applications
ELISA (Assay Dependent), Western Blot (0.1 µg/ml)
Alternate names:
Renal carcinoma antigen NY-REN-59; LDH muscle subunit; Cell proliferation-inducing gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517246 349.00
Check your price
  • LDHA Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517183
  • Size 100µL
Applications
Western Blot (1:1000)
Alternate names:
Renal carcinoma antigen NY-REN-59; LDH muscle subunit; Cell proliferation-inducing gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517183 275.00
Check your price
  • LDHB Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517242
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (2 µg), Western Blot (0.1 µg/ml)
Alternate names:
LDH heart subunit; Renal carcinoma antigen NY-REN-46
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517242 349.00
Check your price
  • LDHA Monoclonal Antibody for Western Blot, Flow, IP and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517247
  • Size 100µL
Applications
ELISA (Assay Dependent), Immunoprecipitation (2 µg), Western Blot (0.1 µg/ml)
Alternate names:
Renal carcinoma antigen NY-REN-59; LDH muscle subunit; Cell proliferation-inducing gene 19 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517247 349.00
Check your price
  • LDHAL6B Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Mouse, Human
  • Conjugate
  • Catalog # PA531189
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
LDH6B, LDHAL6, LDHL, L-lactate dehydrogenase A-like 6B, lactate dehydrogenase A-like 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531189 Not Orderable Online
Check your price
  • LDHB Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527505
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Renal carcinoma antigen NY-REN-46; LDH heart subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527505 349.00
Check your price
  • LDHB Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA543141
  • Size 100µl
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1000)
Alternate names:
LDH heart subunit; Renal carcinoma antigen NY-REN-46
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543141 309.00
Check your price
  • LDHC Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Goat, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544076
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
LDH-X; LDH testis subunit; Cancer/testis antigen 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544076 309.00
Check your price
  • LDHC Polyclonal Antibody for Western Blot, IHC and pep-ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518779
  • Size 100ug
Applications
Immunohistochemistry (Assay Dependent), Western Blot (0.03-0.1 ug/ml)
Alternate names:
LDH-X; LDH testis subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518779 349.00
Check your price
  • LDHC Polyclonal Antibody for Western Blot, IHC and pep-ELISA
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518780
  • Size 100ug
Applications
Immunohistochemistry (Assay Dependent), Western Blot (0.03-0.1 ug/ml)
Alternate names:
LDH-X; LDH testis subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518780 349.00
Check your price
  • XDH Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526285
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
XO, XOR, xanthene dehydrogenase, xanthine dehydrogenase/oxidase, xanthine oxidase, xanthine oxidoreductase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526285 349.00
Check your price
  • DHODH Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539133
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Dihydroorotate oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539133 309.00
Check your price
  • UEVLD Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA539521
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
EV and lactate/malate dehydrogenase domain-containing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539521 309.00
Check your price
  • SLC30A4 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539552
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Solute carrier family 30 member 4; Lethal milk protein; Dri 27 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539552 309.00
Check your price
  • HIF1A Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Bovine
  • Conjugate Unconjugated
  • Catalog # 458400
  • Size 100µg
Applications
Western Blot (2 ug/ml)
Alternate names:
HIF-1alpha, HIF1, HIF1-ALPHA, MOP1, PASD8, bHLHe78, ARNT interacting protein, ARNT-interacting protein, HIF-1-alpha, PAS domain-containing protein 8, basic-helix-loop-helix-PAS protein MOP1, class E basic helix-loop-helix protein 78, hypoxia-inducible factor 1 alpha isoform I.3, hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, member of PAS protein 1, member of PAS superfamily 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
458400 309.00
Check your price