1 - 21 of 21 products displayed
  • Lamin B2 Monoclonal Antibody for Western Blot, IF, IHC (F) and Flow
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Pig, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA106104
  • Size 100 µL
Applications
Flow Cytometry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100- 1:200), Western Blot (1:100-1:1000)
Alternate names:
LAMB2, LMN2, MGC2721
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106104 309.00
Check your price
  • Lamin B2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513697
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Lamin-B2, LMN2; LMNB2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513697 349.00
Check your price
  • Lamin B2 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517274
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
LAMB2, LMN2, lamin-B2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517274 349.00
Check your price
  • Lamin B2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529121
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
LAMB2, LMN2, lamin-B2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529121 349.00
Check your price
  • Lamin B2 Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522066
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
LAMB2, LMN2, lamin-B2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522066 349.00
Check your price
  • Lamin B2 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 332100
  • Size 100 µg
Applications
Immunofluorescence (10 ug/ml), Immunoprecipitation (5 ug), Western Blot (3.0 ug/ml)
Alternate names:
LAMB2, LMN2, lamin-B2
Catalog # Size Price (AUD) Your Price (AUD) Qty
332100 309.00
Check your price
  • Lamin B1/B2 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA190041
  • Size 1 mL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Neat), Western Blot (1:50)
Alternate names:
Lamin-B1, LMNB1, LMN2, LMNB, Lamin-B2, LMNB2, LMN2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190041 399.00
Check your price
  • Lamin B1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA106103
  • Size 100 µL
Applications
ELISA (Assay Dependent), Flow Cytometry (1:100-1:200), Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
ADLD, LMN, LMN2, LMNB, MGC111419
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106103 375.00
Check your price
  • Lamin B1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141074
  • Size 100 µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
ADLD, LMN, LMN2, LMNB, MGC111419
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141074 349.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # MA106102
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
RP11-54H19.1, CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LDP1, LFP, LGMD1B, LMN1, LMNC, PRO1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106102 375.00
Check your price
  • Lamin B1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532474
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (1:25)
Alternate names:
ADLD, LMN, LMN2, LMNB, lamin-B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532474 349.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518073
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:500)
Alternate names:
Prelamin-A/C; 70 kDa lamin; LMN1; LMNA; Renal carcinoma antigen NY-REN-32; Mutant lamin A/C; progerin mutant; Lamin AC; Lamin A/C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518073 425.00
Check your price
  • Lamin A/C (mutant 453W) Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518063
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:32,000)
Alternate names:
Prelamin-A/C; 70 kDa lamin; LMN1; LMNA; Renal carcinoma antigen NY-REN-32; mutant 453W; Lamin A+C mutant
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518063 399.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA15820
  • Size 1 mL
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (1:50)
Alternate names:
Prelamin-A/C, Lamin-A/C, 70 kDa lamin, Renal carcinoma antigen NY-REN-32, LMNA, LMN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15820 475.00
Check your price
  • Lamin A/C (mutant 453W) Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518058
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:8000)
Alternate names:
Prelamin-A/C; 70 kDa lamin; LMN1; LMNA; Renal carcinoma antigen NY-REN-32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518058 399.00
Check your price
  • Lamin B1 Monoclonal Antibody for Western Blot, IF, Flow, IP and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 332000
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunofluorescence (5-10 ug/ml), Immunoprecipitation (2.5-5 ug), Western Blot (0.5-1.0 ug/ml)
Alternate names:
ADLD, LMN, LMN2, LMNB, lamin-B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
332000 305.00
Check your price
  • Lamin C Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA15827
  • Size 200 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:100), Western Blot (Assay Dependent)
Alternate names:
C type lamin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15827 550.00
Check your price
  • Lamin A Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA106101
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106101 309.00
Check your price
  • Lamin B1 Monoclonal Antibody for Western Blot and IF
  • Host Mouse
  • Target Species Bovine, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA15822
  • Size 2 mL
Applications
Immunofluorescence (Assay Dependent), Western Blot (1:50)
Alternate names:
B type lamin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15822 550.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA15821
  • Size 1 mL
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (1:50)
Alternate names:
Prelamin-A/C, Lamin-A/C, 70 kDa lamin, Renal carcinoma antigen NY-REN-32, LMNA, LMN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15821 425.00
Check your price
  • Lamin C Polyclonal Antibody for Western Blot, IHC (F) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA106105
  • Size 100µL
Applications
Flow Cytometry (1:50-1:250), Immunohistochemistry (Frozen) (1:50-1:250), Western Blot (1:50-1:500)
Alternate names:
OR19-15, OR19-8, OR7G1P
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA106105 425.00
Check your price