1 - 30 of 30 products displayed
  • Lamin B2 Monoclonal Antibody for Western Blot, IF, IHC (F) and Flow
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Pig, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA106104
  • Size 100µL
Applications
Flow Cytometry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100- 1:200), Western Blot (1:100-1:1000)
Alternate names:
LAMB2, LMN2, MGC2721
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106104 309.00
Check your price
  • Lamin B1/B2 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA190041
  • Size 1mL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Neat), Western Blot (1:50)
Alternate names:
Lamin-B1, LMNB1, LMN2, LMNB, Lamin-B2, LMNB2, LMN2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190041 399.00
Check your price
  • Lamin B1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA106103
  • Size 100µL
Applications
ELISA (Assay Dependent), Flow Cytometry (1:100-1:200), Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
ADLD, LMN, LMN2, LMNB, MGC111419
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106103 375.00
Check your price
  • Lamin B2 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517274
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
LAMB2, LMN2, lamin-B2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517274 349.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA31000
  • Size 200µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (1:100), Western Blot (1:100-1:2000)
Alternate names:
70 kDa lamin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA31000 225.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # MA106102
  • Size 100µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
RP11-54H19.1, CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LDP1, LFP, LGMD1B, LMN1, LMNC, PRO1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106102 375.00
Check your price
  • LAP1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1074
  • Size 200µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:20-1:50), Immunoprecipitation (Assay dependent), Western Blot (1:200)
Alternate names:
Lamina-associated polypeptide 1B; Lamina-associated polypeptide 1C; Lamin-associated protein 1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1074 275.00
Check your price
  • Lamin B2 Monoclonal Antibody for Western Blot, IF and IP
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 332100
  • Size 100µg
Applications
Immunofluorescence (10 ug/ml), Immunoprecipitation (5 ug), Western Blot (3.0 ug/ml)
Alternate names:
LAMB2, LMN2, lamin-B2
Catalog # Size Price (AUD) Your Price (AUD) Qty
332100 309.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515882
  • Size 100µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
RP11-54H19.1, CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, FPLD2, HGPS, IDC, LDP1, LFP, LGMD1B, LMN1, LMNC, LMNL1, PRO1, 70 kDa lamin; lamin A/C-like 1; lamin-A/C; prelamin-A/C; renal carcinoma antigen NY-REN-32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515882 349.00
Check your price
  • Lamin A (Cleaved Asp230) Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515211
  • Size 100µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:1000)
Alternate names:
70 kDa lamin; OTTHUMP00000015843; OTTHUMP00000015846; OTTHUMP00000015847; lamin A/C-like 1; lamin-A/C; prelamin-A/C; renal carcinoma antigen NY-REN-32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515211 309.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518073
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:500)
Alternate names:
Prelamin-A/C; 70 kDa lamin; LMN1; LMNA; Renal carcinoma antigen NY-REN-32; Mutant lamin A/C; progerin mutant; Lamin AC; Lamin A/C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518073 425.00
Check your price
  • Lamin A/C (mutant 453W) Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518063
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:32,000)
Alternate names:
Prelamin-A/C; 70 kDa lamin; LMN1; LMNA; Renal carcinoma antigen NY-REN-32; mutant 453W; Lamin A+C mutant
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518063 399.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA15820
  • Size 1mL
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (1:50)
Alternate names:
Prelamin-A/C, Lamin-A/C, 70 kDa lamin, Renal carcinoma antigen NY-REN-32, LMNA, LMN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15820 475.00
Check your price
  • Lamin A/C (mutant 453W) Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518058
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:8000)
Alternate names:
Prelamin-A/C; 70 kDa lamin; LMN1; LMNA; Renal carcinoma antigen NY-REN-32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518058 399.00
Check your price
  • Lamin B1 Monoclonal Antibody for Western Blot, IF, Flow, IP and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 332000
  • Size 100µg
Applications
ELISA (0.1-1.0 ug/ml), Immunofluorescence (5-10 ug/ml), Immunoprecipitation (2.5-5 ug), Western Blot (0.5-1.0 ug/ml)
Alternate names:
ADLD, LMN, LMN2, LMNB, lamin-B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
332000 305.00
Check your price
  • Lamin A Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA106101
  • Size 100µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106101 309.00
Check your price
  • Lamin C Monoclonal Antibody for Western Blot, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA110397
  • Size 100ug
Applications
Immunocytochemistry (Assay Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
OR19-15, OR19-8, OR7G1P, olfactory receptor 19-15, olfactory receptor 7G1, olfactory receptor OR19-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110397 225.00
Check your price
  • LAP2 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA125323
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (2-4 µg/ml)
Alternate names:
Thymopoietin isoforms alpha/zeta; Thymopoietin isoform beta; Thymopoietin isoforms beta/delta/epsilon/gamma; Thymopoietin-related peptide isoform alpha; Thymopoietin isoform alpha; Thymopoietin-related peptide isoforms beta/gamma; Thymopoietin, isoforms beta/gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125323 399.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA15821
  • Size 1mL
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (1:50)
Alternate names:
Prelamin-A/C, Lamin-A/C, 70 kDa lamin, Renal carcinoma antigen NY-REN-32, LMNA, LMN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15821 425.00
Check your price
  • Lamin B1 Monoclonal Antibody for Western Blot and IF
  • Host Mouse
  • Target Species Bovine, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA15822
  • Size 2mL
Applications
Immunofluorescence (Assay Dependent), Western Blot (1:50)
Alternate names:
B type lamin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15822 550.00
Check your price
  • PrP Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA110152
  • Size 100ug
Applications
Western Blot (0.5 ug/ml)
Alternate names:
AltPrP; PrP33-35C; PrP27-30; ASCR
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110152 275.00
Check your price
  • Nesprin 1 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA518077
  • Size 100µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Western Blot (1:400-1:800)
Alternate names:
Synaptic nuclear envelope protein 1; Nuclear envelope spectrin repeat protein 1; Myocyte nuclear envelope protein 1; Enaptin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518077 425.00
Check your price
  • PP1 gamma Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517250
  • Size 100µL
Applications
ELISA (Assay Dependent), Western Blot (0.1 µg/ml)
Alternate names:
Protein phosphatase 1C catalytic subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517250 349.00
Check your price
  • BNIP3L Monoclonal Antibody for Western Blot, IF, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 393300
  • Size 100µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
NIP3-like protein X; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A; Adenovirus E1B19K-binding protein B5
Catalog # Size Price (AUD) Your Price (AUD) Qty
393300 304.00
Check your price
  • hnRNP K Monoclonal Antibody for Western Blot, IF, IHC and Flow
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA1087
  • Size 100µg
Applications
Flow Cytometry (1µg per 10^6 cells), Immunofluorescence (5 µg/ml), Immunohistochemistry (5 ug/mL), Western Blot (2 ug/mL)
Alternate names:
dC stretch-binding protein; Transformation up-regulated nuclear protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1087 275.00
Check your price
  • Nesprin 1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA113082X
  • Size 20µL
Applications
Western Blot (1:1000)
Alternate names:
candidate plasticity gene 2, syne-1 fragment
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA113082X 119.00
Check your price
  • Nesprin 1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA113082
  • Size 100µg
Applications
Western Blot (1:1000)
Alternate names:
candidate plasticity gene 2, syne-1 fragment
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA113082 309.00
Check your price
  • PKP1 Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA518082
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (Assay-dependent), Immunohistochemistry (Paraffin) (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:100)
Alternate names:
Band 6 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518082 425.00
Check your price
  • PKP1 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 325700
  • Size 100µg
Applications
Immunofluorescence (3-10 ug/ml), Immunoprecipitation (2 ug), Western Blot (1-3 ug/ml)
Alternate names:
Band 6 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
325700 309.00
Check your price
  • PrP Monoclonal Antibody for Western Blot, IHC (P) and ELISA
  • Host Mouse
  • Target Species Bovine, Deer, Human, Sheep
  • Conjugate Unconjugated
  • Catalog # MA1750
  • Size 200µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin) (3-5 µg/ml), Western Blot (3-5 µg/ml)
Alternate names:
AltPrP; PrP33-35C; PrP27-30; ASCR; Major scrapie-associated fibril protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1750 349.00
Check your price