1 - 30 of 30 products displayed
  • Laminin alpha-3 Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524246
  • Size 100 µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Nicein subunit alpha; Laminin-7 subunit alpha; Laminin-6 subunit alpha; Laminin-5 subunit alpha; Kalinin subunit alpha; Epiligrin subunit alpha; Epiligrin 170 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524246 275.00
Check your price
  • Laminin beta-3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529161
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Nicein subunit beta; Laminin-5 subunit beta; Laminin B1k chain; Kalinin subunit beta; Kalinin B1 chain; Epiligrin subunit bata
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529161 349.00
Check your price
  • Laminin Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA121194
  • Size 500 µL
Applications
Immunohistochemistry (Frozen) (1:5-1:20), Immunohistochemistry (Paraffin) (1:5-1:20)
Alternate names:
S-laminin subunit alpha; Laminin-3 subunit alpha; Laminin-1 subunit alpha; Laminin A chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121194 425.00
Check your price
  • Laminin beta-3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521514
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Nicein subunit beta; Laminin-5 subunit beta; Kalinin subunit beta; Kalinin B1 chain; Epiligrin subunit bata; Laminin B1k chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521514 349.00
Check your price
  • Laminin gamma-2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522308
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Nicein subunit gamma; Large adhesive scatter factor 140 kDa subunit; Laminin-5 subunit gamma; Laminin B2t chain; Ladsin 140 kDa subunit; Kalinin/nicein/epiligrin 100 kDa subunit; Kalinin subunit gamma; Epiligrin subunit gamma; Cell-scattering factor 140 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522308 349.00
Check your price
  • ITGA2 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate PE
  • Catalog # MA516572
  • Size 100 TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516572 349.00
Check your price
  • ITGA2 Monoclonal Antibody for IHC (F), Flow and FN
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA516571
  • Size 200 ug
Applications
Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunohistochemistry (Frozen) (1:500-1:1000)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516571 375.00
Check your price
  • Laminin alpha-3 Polyclonal Antibody for IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538937
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100)
Alternate names:
Nicein subunit alpha; Laminin-7 subunit alpha; Laminin-6 subunit alpha; Laminin-5 subunit alpha; Kalinin subunit alpha; Epiligrin subunit alpha; Epiligrin 170 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538937 309.00
Check your price
  • ITGA2 Monoclonal Antibody for Western Blot, IF, IHC (F), IHC (P), Flow, IP, ELISA and FN
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180784
  • Size 200 µg
Applications
ELISA (10 ug/ml), Flow Cytometry (30 ug/ml), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (30-40 ug/ml), Immunohistochemistry (Paraffin) (Assay Dependent), Immunoprecipitation (25-50 ug/ml), Western Blot (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180784 399.00
Check your price
  • ITGA2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532880
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532880 349.00
Check your price
  • ITGA2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532879
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532879 349.00
Check your price
  • ITGA2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526061
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526061 349.00
Check your price
  • ITGA2 Monoclonal Antibody for IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA146035
  • Size 100 µg
Applications
Flow Cytometry (1:10-1:1000), Immunohistochemistry (Frozen) (1:10-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (1:10-1:500)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA146035 309.00
Check your price
  • ITGA2 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Biotin
  • Catalog # MA180244
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180244 325.00
Check your price
  • ITGA2 Monoclonal Antibody for Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA126714
  • Size 50 µg
Applications
ELISA (10 µg/ml), Flow Cytometry (Assay Dependent), Immunoprecipitation (25-50 µg/ml)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126714 309.00
Check your price
  • ITGA2 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # A15431
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15431 214.00
Check your price
  • ITGA2 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # A15432
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
VLA-2 subunit alpha; Platelet membrane glycoprotein Ia; Collagen receptor; CD49 antigen-like family member B
Catalog # Size Price (AUD) Your Price (AUD) Qty
A15432 239.00
Check your price
  • Laminin alpha-5 Monoclonal Antibody for IF, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183286
  • Size 50 µL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
Laminin-15 subunit alpha; Laminin-11 subunit alpha; Laminin-10 subunit alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183286 650.00
Check your price
  • ITGB5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137931
  • Size 7 mL
Applications
Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
CD61, GP3A, GPIIIa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137931 275.00
Check your price
  • CD49a Polyclonal Antibody, Alexa Fluor® 488 conjugate for Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Alexa Fluor® 488
  • Catalog # PA546887
  • Size 100 Tests
Applications
Flow Cytometry (5 µl/10^6 cells)
Alternate names:
VLA-1; Laminin and collagen receptor; CD49 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546887 249.00
Check your price
  • CD49a Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547763
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
VLA-1; Laminin and collagen receptor; CD49 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547763 349.00
Check your price
  • Lamin B1 Monoclonal Antibody for Western Blot, IF, Flow, IP and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 332000
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunofluorescence (5-10 ug/ml), Immunoprecipitation (2.5-5 ug), Western Blot (0.5-1.0 ug/ml)
Alternate names:
ADLD, LMN, LMN2, LMNB, lamin-B1
Catalog # Size Price (AUD) Your Price (AUD) Qty
332000 305.00
Check your price
  • Laminin gamma-1 Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA519253
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
S-laminin subunit gamma; Laminin-9 subunit gamma; Laminin-8 subunit gamma; Laminin-7 subunit gamma; Laminin-6 subunit gamma; Laminin-4 subunit gamma; Laminin-3 subunit gamma; Laminin-2 subunit gamma; Laminin-11 subunit gamma; Laminin-10 subunit gamma; Laminin-1 subunit gamma; Laminin B2 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA519253 309.00
Check your price
  • RPSA Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA512840
  • Size 500 µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (4-8 µg/mL), Western Blot (1:10-1:200)
Alternate names:
NEM/1CHD4; Multidrug resistance-associated protein MGr1-Ag; Laminin-binding protein precursor p40; Laminin receptor 1; Colon carcinoma laminin-binding protein; 67 kDa laminin receptor; 37/67 kDa laminin receptor; 37 kDa laminin receptor precursor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512840 349.00
Check your price
  • Galectin 1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 437400
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:20), Western Blot (Assay Dependent)
Alternate names:
S-Lac lectin 1; Lectin galactoside-binding soluble 1; Lactose-binding lectin 1; Galaptin; Beta-galactoside-binding lectin L-14-I; 14 kDa lectin; RL 14.5; Putative MAPK-activating protein PM12; HPL; HBL; 14 kDa laminin-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
437400 304.00
Check your price
  • Galectin 3 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1940
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (6 µg/ml), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1 µg/ml)
Alternate names:
Mac-2 antigen; Lectin L-29; Laminin-binding protein; IgE-binding protein; Galactose-specific lectin 3; Carbohydrate-binding protein 35; 35 kDa lectin; L-31; Galactoside-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1940 375.00
Check your price
  • ITGA3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533859
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (2.5-5 µg/ml)
Alternate names:
VLA-3 subunit alpha; Galactoprotein B3; FRP-2; CD49 antigen-like family member C
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533859 349.00
Check your price
  • CD49a Polyclonal Antibody, PE conjugate for Flow
  • Host Sheep
  • Target Species Human
  • Conjugate PE
  • Catalog # PA546890
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
VLA-1; Laminin and collagen receptor; CD49 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546890 279.00
Check your price
  • CD49a Polyclonal Antibody, APC conjugate for Flow
  • Host Sheep
  • Target Species Human
  • Conjugate APC
  • Catalog # PA546899
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
VLA-1; Laminin and collagen receptor; CD49 antigen-like family member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546899 309.00
Check your price
  • CD56 Monoclonal Antibody for Western Blot, IHC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 075603
  • Size 100 µg
Applications
Immunohistochemistry (Assay Dependent), Immunoprecipitation (2-5 µg), Western Blot (Assay Dependent)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
075603 268.00
Check your price