1 - 30 of 30 products displayed
  • LARGE Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519292
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Acetylglucosaminyltransferase-like 1A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519292 349.00
Check your price
  • ARHGEF12 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539418
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Leukemia-associated RhoGEF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539418 309.00
Check your price
  • KCNMB3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533873
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
Slo-beta-3; Maxi K channel subunit beta-3; K(VCA)beta-3; Charybdotoxin receptor subunit beta-3; Calcium-activated potassium channel, subfamily M subunit beta-3; BK channel subunit beta-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533873 349.00
Check your price
  • RFC1 Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535965
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
Replication factor C large subunit; Replication factor C 140 kDa subunit; DNA-binding protein PO-GA; Activator 1 subunit 1; Activator 1 large subunit; Activator 1 140 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535965 309.00
Check your price
  • RFC1 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535966
  • Size 100 µL
Applications
Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
Replication factor C large subunit; Replication factor C 140 kDa subunit; DNA-binding protein PO-GA; Activator 1 subunit 1; Activator 1 large subunit; Activator 1 140 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535966 309.00
Check your price
  • SLC7A5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533053
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (20 µg/ml)
Alternate names:
y+ system cationic amino acid transporter; Solute carrier family 7 member 5; L-type amino acid transporter 1; Integral membrane protein E16; CD98 light chain; 4F2 light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533053 349.00
Check your price
  • SLC7A5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533052
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (13-26 µg/ml)
Alternate names:
y+ system cationic amino acid transporter; Solute carrier family 7 member 5; L-type amino acid transporter 1; Integral membrane protein E16; CD98 light chain; 4F2 light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533052 349.00
Check your price
  • SLC7A5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534216
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (15 µg/ml)
Alternate names:
y+ system cationic amino acid transporter; Solute carrier family 7 member 5; L-type amino acid transporter 1; Integral membrane protein E16; CD98 light chain; 4F2 light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534216 349.00
Check your price
  • SLC7A5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534215
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (7 µg/ml)
Alternate names:
y+ system cationic amino acid transporter; Solute carrier family 7 member 5; L-type amino acid transporter 1; Integral membrane protein E16; CD98 light chain; 4F2 light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534215 349.00
Check your price
  • SLC7A5 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519101
  • Size 100 ug
Applications
Western Blot (1-3 ug/ml)
Alternate names:
y+ system cationic amino acid transporter; Solute carrier family 7 member 5; L-type amino acid transporter 1; Integral membrane protein E16; CD98 light chain; 4F2 light chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519101 349.00
Check your price
  • Calpain 2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA527720
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Millimolar-calpain; Calpain-2 large subunit; Calpain M-type; Calcium-activated neutral proteinase 2; 80 kDa M-calpain subunit; Calpain large polypeptide L2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527720 349.00
Check your price
  • Calpain 2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529381
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Millimolar-calpain; Calpain-2 large subunit; Calpain M-type; Calcium-activated neutral proteinase 2; 80 kDa M-calpain subunit; Calpain large polypeptide L2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529381 349.00
Check your price
  • PSMB6 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Bovine, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1978
  • Size 100 µg
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Western Blot (1 µg/ml)
Alternate names:
Proteasome subunit Y; Proteasome delta chain; Multicatalytic endopeptidase complex delta chain; Macropain delta chain; Low molecular mass protein 19
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1978 349.00
Check your price
  • Mu-Calpain Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527681
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Micromolar-calpain; Calpain-1 large subunit; Calpain mu-type; Calcium-activated neutral proteinase 1; Cell proliferation-inducing gene 30 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527681 349.00
Check your price
  • Mu-Calpain Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527682
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Micromolar-calpain; Cell proliferation-inducing gene 30 protein; Calpain-1 large subunit; Calpain mu-type; Calcium-activated neutral proteinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527682 349.00
Check your price
  • Mu-Calpain Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA126476
  • Size 50 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Micromolar-calpain; Calpain-1 large subunit; Calpain mu-type; Calcium-activated neutral proteinase 1; Cell proliferation-inducing gene 30 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126476 349.00
Check your price
  • Mu-Calpain Monoclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F) and Flow
  • Host Mouse
  • Target Species Bovine, Hamster, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3940
  • Size 100 µg
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Western Blot (1:500)
Alternate names:
Micromolar-calpain; Calpain-1 large subunit; Calpain mu-type; Calcium-activated neutral proteinase 1; Cell proliferation-inducing gene 30 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3940 309.00
Check your price
  • Mu-Calpain Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Bovine, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA3941
  • Size 100 µL
Applications
Immunocytochemistry (1:25), Immunofluorescence (1:20), Western Blot (1:1,000)
Alternate names:
Micromolar-calpain; Calpain-1 large subunit; Calpain mu-type; Calcium-activated neutral proteinase 1; Cell proliferation-inducing gene 30 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3941 275.00
Check your price
  • Mu-Calpain Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3942
  • Size 100 µL
Applications
ELISA (Assay dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (1:20), Immunofluorescence (1:20), Immunohistochemistry (Frozen) (1:50), Western Blot (1:500)
Alternate names:
Micromolar-calpain; Calpain-1 large subunit; Calpain mu-type; Calcium-activated neutral proteinase 1; Cell proliferation-inducing gene 30 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3942 275.00
Check your price
  • CPN2 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514235
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Carboxypeptidase N regulatory subunit; Carboxypeptidase N polypeptide 2; Carboxypeptidase N large subunit; Carboxypeptidase N 83 kDa chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514235 349.00
Check your price
  • GPR98 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533792
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (15 µg/ml)
Alternate names:
Very large G-protein coupled receptor 1; Usher syndrome type-2C protein; Monogenic audiogenic seizure susceptibility protein 1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533792 349.00
Check your price
  • GPR98 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533793
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (4-9 µg/ml)
Alternate names:
Very large G-protein coupled receptor 1; Usher syndrome type-2C protein; Monogenic audiogenic seizure susceptibility protein 1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533793 349.00
Check your price
  • GPR98 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533794
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (10-26 µg/ml), Western Blot (Assay Dependent)
Alternate names:
Very large G-protein coupled receptor 1; Usher syndrome type-2C protein; Monogenic audiogenic seizure susceptibility protein 1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533794 349.00
Check your price
  • GPR98 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532847
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml)
Alternate names:
Very large G-protein coupled receptor 1; Usher syndrome type-2C protein; Monogenic audiogenic seizure susceptibility protein 1 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532847 349.00
Check your price
  • GNAS Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519315
  • Size 100 µg
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
G-alpha-8; Adenylate cyclase-stimulating G alpha protein; Extra large alphas protein; Alternative gene product encoded by XL-exon
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519315 349.00
Check your price
  • Versican V0, V1 Neo Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA11748A
  • Size 100 µg
Applications
Western Blot (1 ug/ml)
Alternate names:
PG-M; Large fibroblast proteoglycan; Glial hyaluronate-binding protein; Chondroitin sulfate proteoglycan core protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11748A 375.00
Check your price
  • SLC43A1 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537059
  • Size 100 µl
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Solute carrier family 43 member 1; Prostate cancer overexpressed gene 1 protein; L-type amino acid transporter 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537059 309.00
Check your price
  • RNaseH2A Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520667
  • Size 100 ug
Applications
Immunocytochemistry (2 µg/ml), Immunofluorescence (4 ug/ml), Western Blot (1 µg/ml)
Alternate names:
Ribonuclease HI subunit A; Ribonuclease HI large subunit; RNase H(35); Aicardi-Goutieres syndrome 4 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520667 349.00
Check your price
  • CD98 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA119509
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Solute carrier family 3 member 2; Lymphocyte activation antigen 4F2 large subunit; 4F2 heavy chain antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119509 225.00
Check your price
  • CD98 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA119586
  • Size 100 tests
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Solute carrier family 3 member 2; Lymphocyte activation antigen 4F2 large subunit; 4F2 heavy chain antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119586 225.00
Check your price