1 - 5 of 5 products displayed
  • CD31 Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate APC
  • Catalog # MA516769
  • Size 100TESTS
Applications
Flow Cytometry (Neat-1:10)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516769 399.00
Check your price
  • CD31 Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate PE
  • Catalog # MA516771
  • Size 100TESTS
Applications
Flow Cytometry (Neat)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516771 550.00
Check your price
  • CD31 Monoclonal Antibody, Biotin conjugate for Flow
  • Host Mouse
  • Target Species Human, Pig
  • Conjugate Biotin
  • Catalog # MA516770
  • Size 100ug
Applications
Flow Cytometry (Neat-1:200)
Alternate names:
PECA1; GPIIA'; EndoCAM
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516770 475.00
Check your price
  • CD31 Monoclonal Antibody for IHC (F) and Flow
  • Host Mouse
  • Target Species Pig
  • Conjugate Unconjugated
  • Catalog # MA182381
  • Size 2mL
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
CD31 antigen, CD31/EndoCAM, GPIIA', PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182381 375.00
Check your price
  • E-cadherin Monoclonal Antibody for IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA514458
  • Size 1mL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Uvomorulin; Epithelial cadherin; CAM 120/80
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514458 550.00
Check your price