1 - 6 of 6 products displayed
  • LRIG1 Polyclonal Antibody for IHC (P) and Flow
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547928
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunohistochemistry (Paraffin) (5-15 µg/ml)
Alternate names:
DKFZp586O1624, LIG-1, LIG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547928 349.00
Check your price
  • LRIG1 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547009
  • Size 100µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (2 µg/ml)
Alternate names:
DKFZp586O1624, LIG-1, LIG1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547009 399.00
Check your price
  • GPR125 Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518934
  • Size 100ug
Applications
Immunohistochemistry (0.1-1.0ug/ml), Western Blot (0.5-1.5 ug/ml)
Alternate names:
G-protein coupled receptor 125
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518934 349.00
Check your price
  • TEM5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532794
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (11 µg/ml)
Alternate names:
Tumor endothelial marker 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532794 349.00
Check your price
  • TEM5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533619
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (2-5 µg/ml)
Alternate names:
Tumor endothelial marker 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533619 349.00
Check your price
  • TEM5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533618
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (4 µg/ml)
Alternate names:
Tumor endothelial marker 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533618 349.00
Check your price