1 - 2 of 2 products displayed
  • LHX3 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA540754
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (1.25 µg/ml)
Alternate names:
Homeobox protein P-LIM; Homeobox protein LIM-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540754 309.00
Check your price
  • LHX3 Polyclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and IP
  • Host Rabbit
  • Target Species Amphibian, Fish, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129491
  • Size 50µg
Applications
Immunocytochemistry (1:100-1:250), Immunofluorescence (1:100-250), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:125-1:500), Immunoprecipitation (1:100), Western Blot (1:1250-1:2500)
Alternate names:
Homeobox protein P-LIM; Homeobox protein LIM-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129491 425.00
Check your price