1 - 5 of 5 products displayed
  • LMO4 Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA524248
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
LIM domain only protein 4; Breast tumor autoantigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524248 349.00
Check your price
  • LMO4 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA125725
  • Size 50 µg
Applications
Western Blot (1:2,000)
Alternate names:
LIM domain only protein 4; Breast tumor autoantigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125725 425.00
Check your price
  • CSRP3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529155
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Muscle LIM protein; LIM domain protein, cardiac; Cysteine-rich protein 3; Cardiac LIM protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529155 349.00
Check your price
  • CSRP3 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA120849
  • Size 50 µg
Applications
Western Blot (1/500)
Alternate names:
Muscle LIM protein; LIM domain protein, cardiac; Cysteine-rich protein 3; Cardiac LIM protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA120849 449.00
Check your price
  • CSRP3 Polyclonal Antibody for Western Blot, ELISA and pep-ELISA
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519062
  • Size 100 ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
Muscle LIM protein; LIM domain protein, cardiac; Cysteine-rich protein 3; Cardiac LIM protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519062 349.00
Check your price