1 - 21 of 21 products displayed
  • LIN28A Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515453
  • Size 100 µL
Applications
Immunocytochemistry (1/200-1/1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515453 349.00
Check your price
  • LIN28A Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA523408
  • Size 100 ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml), Western Blot (3-6 µg/ml)
Alternate names:
Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523408 349.00
Check your price
  • LIN28A Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520898
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520898 349.00
Check your price
  • LIN28A Monoclonal Antibody, DyLight® 650 conjugate for IF, ICC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # MA1016D650
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:500), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100)
Alternate names:
Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1016D650 275.00
Check your price
  • LIN28A Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1096X
  • Size 20 µL
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:20-1:50), Immunoprecipitation (5 µg, 1:1000), Western Blot (1:1000)
Alternate names:
Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1096X 119.00
Check your price
  • LIN28A Monoclonal Antibody, DyLight® 488 conjugate for IF, ICC and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # MA1016D488
  • Size 100 µL
Applications
Flow Cytometry (1:50-1:500), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500)
Alternate names:
Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1016D488 257.00
Check your price
  • LIN28A Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow, IP and ChIP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA1016
  • Size 100 µg
Applications
ChIP assay (1-3 ul), Flow Cytometry (1:100), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (5 µg), Western Blot (1:1000)
Alternate names:
Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1016 249.00
Check your price
  • LIN28A Monoclonal Antibody, DyLight® 550 conjugate for IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate DyLight® 550
  • Catalog # MA1016D550
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100)
Alternate names:
Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1016D550 257.00
Check your price
  • LIN28A Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate HRP
  • Catalog # MA1016HRP
  • Size 100 µL
Applications
Western Blot (1:250-1:1000 )
Alternate names:
Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1016HRP 275.00
Check your price
  • LIN28A Polyclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1096
  • Size 100 µg
Applications
Flow Cytometry (1:100), Immunocytochemistry (1:100-1:200), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin) (1:20-1:50), Immunoprecipitation (5 µg, 1:1000), Western Blot (1:1000)
Alternate names:
Testis-expressed protein 17; Zinc finger CCHC domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1096 349.00
Check your price
  • SOX2 Monoclonal Antibody, DyLight® 650 conjugate for IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate DyLight® 650
  • Catalog # MA1014D650
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100)
Alternate names:
ANOP3, MCOPS3, SRY-related HMG-box gene 2, transcription factor SOX-2, transcription factor SOX2, sex determining region Y box 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1014D650 275.00
Check your price
  • Nanog Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520889
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Homeobox transcription factor Nanog; Early embryo specific expression NK-type homeobox protein; ES cell-associated protein 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520889 349.00
Check your price
  • OCT4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520887
  • Size 100 ug
Applications
Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Octamer-binding transcription factor 3; Octamer-binding protein 4; Octamer-binding protein 3; NF-A3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520887 349.00
Check your price
  • KLF4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520897
  • Size 100 ug
Applications
Immunohistochemistry (5 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Gut-enriched krueppel-like factor; Epithelial zinc finger protein EZF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520897 349.00
Check your price
  • SOX2 Monoclonal Antibody, DyLight® 488 conjugate for IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate DyLight® 488
  • Catalog # MA1014D488
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100)
Alternate names:
ANOP3, MCOPS3, SRY-related HMG-box gene 2, transcription factor SOX-2, transcription factor SOX2, sex determining region Y box 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1014D488 257.00
Check your price
  • OCT4 Monoclonal Antibody for Western Blot, IF and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # A13998
  • Size 100 µL
Applications
Flow Cytometry (1:600), Immunofluorescence (1:400), Western Blot (1:1,000)
Alternate names:
Octamer-binding transcription factor 3; Octamer-binding protein 4; Octamer-binding protein 3; NF-A3
Catalog # Size Price (AUD) Your Price (AUD) Qty
A13998 306.00
Check your price
  • Nanog Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Goat
  • Target Species Human, Pig
  • Conjugate Unconjugated
  • Catalog # PA518406
  • Size 100 ug
Applications
Immunocytochemistry (1-10ug/ml), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (0.03-0.1 ug/ml)
Alternate names:
Homeobox transcription factor Nanog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518406 349.00
Check your price
  • Nanog Polyclonal Antibody for Western Blot, IHC and ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518678
  • Size 100 ug
Applications
ELISA (5-10ug/ml), Immunohistochemistry (1-2µg/ml), Western Blot (0.05-0.1 ug/ml)
Alternate names:
Homeobox transcription factor Nanog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518678 349.00
Check your price
  • OCT4 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Human, Mouse
  • Conjugate PE
  • Catalog # A18525
  • Size 25µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
Octamer-binding transcription factor 3; Octamer-binding protein 4; Octamer-binding protein 3; NF-A3
Catalog # Size Price (AUD) Your Price (AUD) Qty
A18525 119.00
Check your price
  • Mouse IgG2a Isotype Control, DyLight® 650 conjugate for IF, ICC and Flow
  • Host Mouse
  • Target Species Not Applicable
  • Conjugate DyLight® 650
  • Catalog # MA1193D650
  • Size 100 µg
Applications
Flow Cytometry (10µg/ml), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100)
Alternate names:
Negative Control, Immunoglobulin Control, Control
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1193D650 199.00
Check your price
  • Mouse IgG2a Isotype Control, DyLight® 488 conjugate for IF, ICC and Flow
  • Host Mouse
  • Target Species Not Applicable
  • Conjugate DyLight® 488
  • Catalog # MA1193D488
  • Size 100 µg
Applications
Flow Cytometry (10µg/ml), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100)
Alternate names:
Negative Control, Immunoglobulin Control, Control
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1193D488 199.00
Check your price