1 - 2 of 2 products displayed
  • LIAS Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA522387
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Lipoic acid synthase; Lipoate synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522387 349.00
Check your price
  • LIAS Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA543211
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Lipoic acid synthase; Lipoate synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA543211 309.00
Check your price