1 - 30 of 30 products displayed
  • Folate Receptor alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542004
  • Size 100 µl
Applications
Immunocytochemistry (1:100-1:600), Immunofluorescence (1:100-1:600), Immunohistochemistry (Paraffin) (1:100-1:600), Western Blot (0.2-1 µg/ml)
Alternate names:
Ovarian tumor-associated antigen MOv18; KB cells FBP; Folate receptor, adult; Folate receptor 1; Adult folate-binding protein; Milk folate-binding protein; Folate-binding protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542004 309.00
Check your price
  • LXR alpha/beta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1331B
  • Size 200 µg
Applications
Western Blot (1:500)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha; Ubiquitously-expressed nuclear receptor; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor NER; Liver X receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1331B 349.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1330
  • Size 200 µg
Applications
Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Western Blot (1:300)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1330 375.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529868
  • Size 100 µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529868 349.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528439
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528439 349.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA1332
  • Size 100 µg
Applications
Western Blot (1:500)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1332 375.00
Check your price
  • LXR alpha Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # 418300
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
418300 426.00
Check your price
  • LXR alpha Monoclonal Antibody for Western Blot, IP, ELISA and GS
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 418200
  • Size 100 µg
Applications
ELISA (Assay Dependent), Gel Shift (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
418200 433.00
Check your price
  • PPAR alpha Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 424600
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:500), Western Blot (1-3 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
424600 309.00
Check your price
  • THRA Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA121134
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
c-erbA-alpha; c-erbA-1; Nuclear receptor subfamily 1 group A member 1; V-erbA-related protein 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121134 349.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520880
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520880 349.00
Check your price
  • LXR alpha/beta Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19066
  • Size 100 ug
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha; Ubiquitously-expressed nuclear receptor; Retinoid X receptor-interacting protein No.15; Nuclear receptor subfamily 1 group H member 2; Liver X receptor beta; Nuclear receptor NER; Orphan nuclear receptor OR-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19066 349.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Pig
  • Conjugate Unconjugated
  • Catalog # PA116824
  • Size 100 µL
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116824 349.00
Check your price
  • LXR alpha Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526951
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526951 349.00
Check your price
  • PPAR alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Pig, Rat, Sheep, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA132484
  • Size 100 µg
Applications
Western Blot (3-6 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132484 499.00
Check your price
  • F1A alpha Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519955
  • Size 100 ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
FEM1-beta; Fem-1-like in apoptotic pathway protein alpha; Fem-1-like death receptor-binding protein alpha; mt-Fem
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519955 349.00
Check your price
  • MIP-3 alpha Monoclonal Antibody for Western Blot
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516253
  • Size 100 ug
Applications
Western Blot (0.1-2 µg/ml)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1; CC chemokine ST38
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516253 349.00
Check your price
  • F1A alpha Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA130706
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1:500-1:1000)
Alternate names:
FEM1-beta; Fem-1-like in apoptotic pathway protein alpha; Fem-1-like death receptor-binding protein alpha; mt-Fem
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA130706 449.00
Check your price
  • TNF alpha Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA519810
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Tumor necrosis factor ligand superfamily member 2; TNF-alpha; Cachectin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519810 349.00
Check your price
  • LRP1 Monoclonal Antibody for Western Blot, IF, IHC (F), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180192
  • Size 200 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1/50-1/100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay dependent (Non-redu)
Alternate names:
Apolipoprotein E receptor; Alpha-2-macroglobulin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180192 499.00
Check your price
  • PPAR alpha Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA186844
  • Size 50 µg
Applications
ELISA (1:8000-1:32,000), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 1 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA186844 375.00
Check your price
  • MIP-3 alpha Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA523684
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523684 359.00
Check your price
  • MIP-3 alpha Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523583
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523583 279.00
Check your price
  • MIP-3 alpha Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523563
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523563 249.00
Check your price
  • MIP-3 alpha Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547517
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (0.2-1.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547517 349.00
Check your price
  • MIP-3 alpha Monoclonal Antibody for Western Blot, Flow, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523843
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Neutralization (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523843 275.00
Check your price
  • THRA Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529618
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
c-erbA-alpha; c-erbA-1; V-erbA-related protein 7; Nuclear receptor subfamily 1 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529618 349.00
Check your price
  • THRA Polyclonal Antibody for Western Blot, IF, ICC and IP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521337
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunoprecipitation (1:100-1:500), Western Blot (1:500-1:3000)
Alternate names:
c-erbA-alpha; c-erbA-1; V-erbA-related protein 7; Nuclear receptor subfamily 1 group A member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521337 349.00
Check your price
  • RARA Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539870
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Nuclear receptor subfamily 1 group B member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539870 309.00
Check your price
  • Phospho-RARA (Ser77) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA540165
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:3000)
Alternate names:
Nuclear receptor subfamily 1 group B member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540165 309.00
Check your price