1 - 30 of 30 products displayed
  • LXR beta Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1333
  • Size 100µg
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1:200-1:1000), Western Blot (1:200-1:2000)
Alternate names:
Ubiquitously-expressed nuclear receptor; Retinoid X receptor-interacting protein No.15; Nuclear receptor subfamily 1 group H member 2; Liver X receptor beta; Nuclear receptor NER; Orphan nuclear receptor OR-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1333 349.00
Check your price
  • LXR beta Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529771
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Ubiquitously-expressed nuclear receptor; Retinoid X receptor-interacting protein No.15; Nuclear receptor subfamily 1 group H member 2; Liver X receptor beta; Nuclear receptor NER
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529771 349.00
Check your price
  • LXR beta Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528127
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Ubiquitously-expressed nuclear receptor; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor NER; Liver X receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528127 349.00
Check your price
  • LXR beta Monoclonal Antibody for Western Blot, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 418400
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Ubiquitously-expressed nuclear receptor; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor NER; Liver X receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
418400 433.00
Check your price
  • Phospho-beta-2 Adrenergic Receptor (Ser346) Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538728
  • Size 100µg
Applications
Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538728 309.00
Check your price
  • LXR alpha/beta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1331B
  • Size 200µg
Applications
Western Blot (1:500)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha; Ubiquitously-expressed nuclear receptor; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor NER; Liver X receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1331B 349.00
Check your price
  • LXR alpha/beta Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA19066
  • Size 100ug
Applications
Western Blot (1-3 µg/ml)
Alternate names:
Nuclear receptor subfamily 1 group H member 3; Liver X receptor alpha; Ubiquitously-expressed nuclear receptor; Retinoid X receptor-interacting protein No.15; Nuclear receptor subfamily 1 group H member 2; Liver X receptor beta; Nuclear receptor NER; Orphan nuclear receptor OR-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA19066 349.00
Check your price
  • LXR beta Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520886
  • Size 100µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Ubiquitously-expressed nuclear receptor; Retinoid X receptor-interacting protein No.15; Nuclear receptor subfamily 1 group H member 2; Liver X receptor beta; Nuclear receptor NER; Orphan nuclear receptor OR-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520886 349.00
Check your price
  • LXR beta Polyclonal Antibody for Western Blot and IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518310
  • Size 100ug
Applications
Immunohistochemistry (1-2µg/ml), Western Blot (1-3 ug/ml)
Alternate names:
Ubiquitously-expressed nuclear receptor; Nuclear receptor subfamily 1 group H member 2; Nuclear receptor NER; Liver X receptor beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518310 349.00
Check your price
  • beta-2 Adrenergic Receptor Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OPA115269
  • Size 25ug
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA115269 Not Orderable Online
Check your price
  • beta-2 Adrenergic Receptor Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA527083
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527083 349.00
Check your price
  • beta-2 Adrenergic Receptor Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA125659
  • Size 50µg
Applications
Western Blot (1:2000)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA125659 425.00
Check your price
  • beta-2 Adrenergic Receptor Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514116
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514116 Not Orderable Online
Check your price
  • C/EBP beta Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1826
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (2 ug/ml)
Alternate names:
Transcription factor 5; Nuclear factor NF-IL6; Liver-enriched inhibitory protein; Liver activator protein; Silencer factor B; Liver-enriched transcriptional activator; Interleukin-6-dependent-binding protein; C/EBP-related protein 2; AGP/EBP
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1826 349.00
Check your price
  • Phospho-beta-2 Adrenergic Receptor (Ser355, Ser356) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA538403
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538403 309.00
Check your price
  • Phospho-C/EBP beta (Thr188, Thr235) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536629
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Transcription factor 5; Nuclear factor NF-IL6; Liver-enriched inhibitory protein; Liver activator protein; Silencer factor B; Liver-enriched transcriptional activator; Interleukin-6-dependent-binding protein; C/EBP-related protein 2; AGP/EBP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536629 349.00
Check your price
  • Phospho-CEBP beta pThr188 + pThr235 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate
  • Catalog # PA535467
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Transcription factor 5; Nuclear factor NF-IL6; Liver-enriched inhibitory protein; Liver activator protein; Silencer factor B; Liver-enriched transcriptional activator; Interleukin-6-dependent-binding protein; C/EBP-related protein 2; AGP/EBP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535467 Not Orderable Online
Check your price
  • C/EBP beta Polyclonal Antibody for Western Blot, IHC (P), IP and ChIP
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527244
  • Size 100µL
Applications
ChIP assay (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Transcription factor 5; Nuclear factor NF-IL6; Liver-enriched inhibitory protein; Liver activator protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527244 349.00
Check your price
  • beta Catenin Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA516429
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Immunoprecipitation (10 µg/ml), Western Blot (5-10 µg/mL)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516429 349.00
Check your price
  • Phospho-beta Catenin (Ser33, Ser37) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 486000
  • Size 100µL
Applications
Western Blot (1:1,000)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
486000 309.00
Check your price
  • Phospho-C/EBP beta (Thr235) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540195
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Transcription factor 5; Nuclear factor NF-IL6; Liver-enriched inhibitory protein; Liver activator protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540195 309.00
Check your price
  • beta Catenin Polyclonal Antibody for Western Blot, IF, IHC (F), IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Human, Mouse, Rodent, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA519469
  • Size 100µg
Applications
ELISA (0.1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Frozen) (1:100), Immunohistochemistry (Paraffin) (1 µg/ml), Western Blot (0.25 µg/ml)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519469 349.00
Check your price
  • beta Catenin Polyclonal Antibody for IF, ICC, IHC (P), IP and ELISA
  • Host Rabbit
  • Target Species Chicken, Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # 712700
  • Size 100µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:20-1:200), Immunoprecipitation (Assay Dependent)
Alternate names:
Beta-catenin
Catalog # Size Price (AUD) Your Price (AUD) Qty
712700 305.00
Check your price
  • RARB Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # P21937
  • Size 100µL
Applications
Western Blot (Assay Dependent)
Alternate names:
RAR-epsilon; Nuclear receptor subfamily 1 group B member 2; HBV-activated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
P21937 Not Orderable Online
Check your price
  • beta-2 Adrenergic Receptor Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # OPA115267
  • Size 25µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
OPA115267 Not Orderable Online
Check your price
  • Phospho-beta-2 Adrenergic Receptor (Ser346) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536784
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536784 349.00
Check your price
  • beta-2 Adrenergic Receptor Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # PA518423
  • Size 100ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518423 349.00
Check your price
  • beta-1,2 Adaptin Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Bovine, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA125065
  • Size 100µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay Dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:200)
Alternate names:
Plasma membrane adaptor HA2/AP2 adaptin beta subunit; Clathrin assembly protein complex 2 beta large chain; Beta-adaptin; Beta-2-adaptin; Adaptor-related protein complex 2 subunit beta; Adaptor protein complex AP-2 subunit beta; AP105B; Golgi adaptor HA1/AP1 adaptin beta subunit; Clathrin assembly protein complex 1 beta large chain; Beta-adaptin 1; Beta-1-adaptin; Adaptor-related protein complex 1 subunit beta-1; Adaptor protein complex AP-1 subunit beta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA125065 349.00
Check your price
  • Phospho-beta-2 Adrenergic Receptor (Ser261) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512977
  • Size 400µL
Applications
Dot blot (1:500)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512977 349.00
Check your price
  • beta-2 Adrenergic Receptor Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533335
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (17-26 µg/ml)
Alternate names:
Beta-2 adrenoreceptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533335 349.00
Check your price