1 - 25 of 25 products displayed
  • Lamin A/C Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA141104
  • Size 200 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
LMNA
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141104 375.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518073
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:500)
Alternate names:
Prelamin-A/C; 70 kDa lamin; LMN1; LMNA; Renal carcinoma antigen NY-REN-32; Mutant lamin A/C; progerin mutant; Lamin AC; Lamin A/C
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518073 425.00
Check your price
  • Lamin A/C (mutant 453W) Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518063
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:32,000)
Alternate names:
Prelamin-A/C; 70 kDa lamin; LMN1; LMNA; Renal carcinoma antigen NY-REN-32; mutant 453W; Lamin A+C mutant
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518063 399.00
Check your price
  • Phospho-Lamin A (Ser22) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517113
  • Size 100 µL
Applications
Immunocytochemistry (1:800), Immunofluorescence (1:800), Western Blot (1:1000)
Alternate names:
Prelamin-A/C, Lamin-A/C, 70 kDa lamin, Renal carcinoma antigen NY-REN-32, LMNA, LMN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517113 309.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA15820
  • Size 1 mL
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (1:50)
Alternate names:
Prelamin-A/C, Lamin-A/C, 70 kDa lamin, Renal carcinoma antigen NY-REN-32, LMNA, LMN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15820 475.00
Check your price
  • Lamin A/C (mutant 453W) Monoclonal Antibody for Western Blot, IF, ICC and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518058
  • Size 100 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunoprecipitation (Assay-dependent), Western Blot (1:8000)
Alternate names:
Prelamin-A/C; 70 kDa lamin; LMN1; LMNA; Renal carcinoma antigen NY-REN-32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518058 399.00
Check your price
  • Phospho-Lamin A (Ser392) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538290
  • Size 100 µg
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Prelamin-A/C, Lamin-A/C, 70 kDa lamin, Renal carcinoma antigen NY-REN-32, LMNA, LMN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538290 309.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA15821
  • Size 1 mL
Applications
Immunocytochemistry (1:10), Immunofluorescence (1:10), Western Blot (1:50)
Alternate names:
Prelamin-A/C, Lamin-A/C, 70 kDa lamin, Renal carcinoma antigen NY-REN-32, LMNA, LMN1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA15821 425.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot and IHC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515882
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
RP11-54H19.1, CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, FPLD2, HGPS, IDC, LDP1, LFP, LGMD1B, LMN1, LMNC, LMNL1, PRO1, 70 kDa lamin; lamin A/C-like 1; lamin-A/C; prelamin-A/C; renal carcinoma antigen NY-REN-32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515882 349.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF, ICC and IHC (F)
  • Host Mouse
  • Target Species Bovine, Human, Pig
  • Conjugate Unconjugated
  • Catalog # MA31000
  • Size 200 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (1:100), Western Blot (1:100-1:2000)
Alternate names:
70 kDa lamin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA31000 225.00
Check your price
  • Lamin A (Cleaved Asp230) Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515211
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Western Blot (1:1000)
Alternate names:
70 kDa lamin; OTTHUMP00000015843; OTTHUMP00000015846; OTTHUMP00000015847; lamin A/C-like 1; lamin-A/C; prelamin-A/C; renal carcinoma antigen NY-REN-32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515211 309.00
Check your price
  • Lamin A (Cleaved Asp230) Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517838
  • Size 100 µL
Applications
Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:100), Western Blot (1:1000)
Alternate names:
70 kDa lamin; OTTHUMP00000015843; OTTHUMP00000015846; OTTHUMP00000015847; lamin A/C-like 1; lamin-A/C; prelamin-A/C; renal carcinoma antigen NY-REN-32
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517838 309.00
Check your price
  • Lamin A/C Monoclonal Antibody for Western Blot, IF, ICC, IHC (F) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Hamster, Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # MA106102
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
RP11-54H19.1, CDCD1, CDDC, CMD1A, CMT2B1, EMD2, FPL, FPLD, HGPS, IDC, LDP1, LFP, LGMD1B, LMN1, LMNC, PRO1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106102 375.00
Check your price
  • Lamin C Monoclonal Antibody for Western Blot, ICC and IP
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA110397
  • Size 100 ug
Applications
Immunocytochemistry (Assay Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
OR19-15, OR19-8, OR7G1P, olfactory receptor 19-15, olfactory receptor 7G1, olfactory receptor OR19-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110397 225.00
Check your price
  • Lamin A Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P) and Flow
  • Host Mouse
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA106101
  • Size 100 µg
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Frozen) (1:100-1:200), Immunohistochemistry (Paraffin) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106101 309.00
Check your price
  • Lamin C Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA15827
  • Size 200 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:100), Western Blot (Assay Dependent)
Alternate names:
C type lamin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA15827 550.00
Check your price
  • ZMPSTE24 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA116964
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1.0-2 µg/ml)
Alternate names:
Zinc metalloproteinase Ste24 homolog; Prenyl protein-specific endoprotease 1; Farnesylated proteins-converting enzyme 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116964 349.00
Check your price
  • ZMPSTE24 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA116965
  • Size 100 µL
Applications
Western Blot (2 µg/ml)
Alternate names:
Zinc metalloproteinase Ste24 homolog; Prenyl protein-specific endoprotease 1; Farnesylated proteins-converting enzyme 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116965 309.00
Check your price
  • Lamin C Polyclonal Antibody for Western Blot, IHC (F) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA106105
  • Size 100µL
Applications
Flow Cytometry (1:50-1:250), Immunohistochemistry (Frozen) (1:50-1:250), Western Blot (1:50-1:500)
Alternate names:
OR19-15, OR19-8, OR7G1P
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA106105 425.00
Check your price
  • ZC3H11A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537081
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Zinc finger CCCH domain-containing protein 11A, KIAA0663, ZC3HDC11A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537081 309.00
Check your price
  • UBAP2L Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536998
  • Size 100 µl
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Protein NICE-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536998 309.00
Check your price
  • LRRC41 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536596
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Protein Muf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536596 309.00
Check your price
  • ZC3H11A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA539539
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Zinc finger CCCH domain-containing protein 11A, KIAA0663, ZC3HDC11A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539539 309.00
Check your price
  • UBAP2L Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539520
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Protein NICE-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539520 309.00
Check your price
  • LRRC41 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539224
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Protein Muf1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539224 309.00
Check your price