1 - 4 of 4 products displayed
  • LMP2 Polyclonal Antibody for IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate
  • Catalog # PA528481
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000)
Alternate names:
Really interesting new gene 12 protein; Proteasome subunit beta-1i; Proteasome chain 7; Multicatalytic endopeptidase complex chain 7; Macropain chain 7; Low molecular mass protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528481 339.00
Check your price
  • PSMB9 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA11960
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Really interesting new gene 12 protein; Proteasome subunit beta-1i; Proteasome chain 7; Multicatalytic endopeptidase complex chain 7; Macropain chain 7; Low molecular mass protein 2; LMP-2d
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11960 375.00
Check your price
  • PSMB9 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519384
  • Size 100 ug
Applications
Western Blot (0.3-1.0 ug/ml)
Alternate names:
Really interesting new gene 12 protein; Proteasome subunit beta-1i; Proteasome chain 7; Multicatalytic endopeptidase complex chain 7; Macropain chain 7; Low molecular mass protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519384 349.00
Check your price
  • Mouse,Human 20S Proteasome LMP2 (residues 207-220) Synthetic Peptide for Ctrl
  • Host N/A
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PEP176
  • Size 50 µg
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP176 109.00
Check your price