1 - 30 of 30 products displayed
  • GAPDH Loading Control Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Bacteria, Dog, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA5157381MG
  • Size 1 mg
Applications
ELISA (Assay Dependent), Flow Cytometry (2 ug/test), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1:1000-1:10,000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA5157381MG 950.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody for Western Blot, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA515739
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (2 ug/test), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000-1:10,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739 225.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, DyLight 800 4X PEG conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight 800 4X PEG
  • Catalog # MA515738D800
  • Size 50 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738D800 225.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, Alexa Fluor® 647 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate Alexa Fluor® 647
  • Catalog # MA515738A647
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (Assay-Dependent)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738A647 225.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, Alexa Fluor® 555 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate Alexa Fluor® 555
  • Catalog # MA515738A555
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (Assay-Dependent)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738A555 225.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, Alexa Fluor® 488 conjugate for Western Blot
  • Host Mouse
  • Target Species Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate Alexa Fluor® 488
  • Catalog # MA515738A488
  • Size 50 µl
Applications
Western Blot (Assay-Dependent)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738A488 225.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, DyLight® 680 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate DyLight® 680
  • Catalog # MA515738D680
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500-1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738D680 159.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, DyLight® 550 conjugate for IF and ICC
  • Host Mouse
  • Target Species Yeast, Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat
  • Conjugate DyLight® 550
  • Catalog # MA515738D550
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738D550 159.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, DyLight® 488 conjugate for IF and ICC
  • Host Mouse
  • Target Species Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate DyLight® 488
  • Catalog # MA515738D488
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738D488 159.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, DyLight® 650 conjugate for IF and ICC
  • Host Mouse
  • Target Species Human, Insect, Mouse, Rabbit, Rat, Yeast, Bacteria, Chicken, Hamster
  • Conjugate DyLight® 650
  • Catalog # MA515738D650
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738D650 159.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate Biotin
  • Catalog # MA515738BTIN
  • Size 50 µL
Applications
ELISA (Assay Dependent), Flow Cytometry (2 ug/test), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1:500-1:1000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738BTIN 159.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate HRP
  • Catalog # MA515738HRP
  • Size 50 µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738HRP 159.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, Alexa Fluor® 647 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Alexa Fluor® 647
  • Catalog # MA516308A647
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308A647 225.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, Alexa Fluor® 555 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Alexa Fluor® 555
  • Catalog # MA516308A555
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308A555 225.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, Alexa Fluor® 488 conjugate for Western Blot
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Alexa Fluor® 488
  • Catalog # MA516308A488
  • Size 50 µl
Applications
Western Blot (1:500)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308A488 225.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, DyLight® 680 conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate DyLight® 680
  • Catalog # MA516308D680
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500-1:1000)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308D680 159.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, DyLight® 550 conjugate for IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate DyLight® 550
  • Catalog # MA516308D550
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308D550 159.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, DyLight® 488 conjugate for IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate DyLight® 488
  • Catalog # MA516308D488
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308D488 159.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, DyLight® 650 conjugate for IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate DyLight® 650
  • Catalog # MA516308D650
  • Size 50 µL
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308D650 159.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, Biotin conjugate for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA516308BTIN
  • Size 50 µL
Applications
ELISA (Assay Dependent), Flow Cytometry (1 ug/test), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308BTIN 159.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, HRP conjugate for Western Blot
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate HRP
  • Catalog # MA516308HRP
  • Size 50 µL
Applications
Western Blot (1:500-1:2000)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308HRP 159.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA516308
  • Size 100 µg
Applications
ELISA (Assay dependent), Flow Cytometry (1 ug/test), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1:2000-1:5000)
Alternate names:
Tubulin beta 4'; Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308 225.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody, DyLight 800 4X PEG conjugate for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate DyLight 800 4X PEG
  • Catalog # MA516308D800
  • Size 50 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516308D800 225.00
Check your price
  • beta Tubulin Loading Control Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Dog, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA5163081MG
  • Size 1 mg
Applications
ELISA (Assay dependent), Flow Cytometry (1 ug/test), Immunocytochemistry (1:50-1:500), Immunofluorescence (1:50-1:500), Immunohistochemistry (Paraffin) (1:10-1:500), Western Blot (1:2000-1:5000)
Alternate names:
Tubulin beta-5 chain
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA5163081MG 950.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody, DyLight 800 4X PEG conjugate for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate DyLight 800 4X PEG
  • Catalog # MA515739D800
  • Size 50 µL
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (1:500-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739D800 159.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody, DyLight® 488 conjugate for IF and ICC
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate DyLight® 488
  • Catalog # MA515739D488
  • Size 50 µL
Applications
Immunocytochemistry (1:20-1:100), Immunofluorescence (1:20-1:100)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739D488 159.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody, DyLight® 680 conjugate for Western Blot
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate DyLight® 680
  • Catalog # MA515739D680
  • Size 50 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739D680 159.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody, Biotin conjugate for Western Blot and Flow
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA515739BTIN
  • Size 50 µL
Applications
Flow Cytometry (2 ug/test), Western Blot (1:500-1:1000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515739BTIN 159.00
Check your price
  • beta Actin Loading Control Monoclonal Antibody for Western Blot, IF, ICC, Flow and ELISA
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA5157391MG
  • Size 1 mg
Applications
ELISA (Assay dependent), Flow Cytometry (1-2 ug/test), Western Blot (1:1000-1:10,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA5157391MG 950.00
Check your price
  • GAPDH Loading Control Monoclonal Antibody for Western Blot, IF, ICC, IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Bacteria, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA515738
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (2 ug/test), Immunocytochemistry (1:20-1:200), Immunofluorescence (1:20-1:200), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1:1000-1:10,000)
Alternate names:
Peptidyl-cysteine S-nitrosylase GAPDH; BARS-38; 38 kDa BFA-dependent ADP-ribosylation substrate
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515738 219.00
Check your price