1 - 30 of 30 products displayed
  • LRP6 Monoclonal Antibody for Western Blot
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523995
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
LRP6, low density lipoprotein receptor-related protein 6, low density lipoprotein-related protein 6, ADCAD2, FLJ90062, FLJ90421
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523995 275.00
Check your price
  • LRP6 Monoclonal Antibody for IF, ICC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523950
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
LRP6, low density lipoprotein receptor-related protein 6, low density lipoprotein-related protein 6, ADCAD2, FLJ90062, FLJ90421
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523950 275.00
Check your price
  • LRP6 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547145
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
LRP6, low density lipoprotein receptor-related protein 6, low density lipoprotein-related protein 6, ADCAD2, FLJ90062, FLJ90421
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547145 375.00
Check your price
  • LRP6 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518317
  • Size 100ug
Applications
Immunohistochemistry (4-6µg/ml)
Alternate names:
LRP6; low density lipoprotein receptor-related protein 6; low density lipoprotein-related protein 6; ADCAD2; FLJ90062; FLJ90421
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518317 349.00
Check your price
  • RAP Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA542711
  • Size 100µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (2.5 µg/ml)
Alternate names:
Low density lipoprotein receptor-related protein-associated protein 1; Heparin-binding protein 44; Gp330-binding 45 kDa protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542711 309.00
Check your price
  • RAP Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527986
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Low density lipoprotein receptor-related protein-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527986 349.00
Check your price
  • RAP Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535336
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
Low density lipoprotein receptor-related protein-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535336 349.00
Check your price
  • CORIN Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Rat, Bovine, Horse, Guinea pig, Human, Mouse, Pig, Rabbit
  • Conjugate Unconjugated
  • Catalog # PA542916
  • Size 100µl
Applications
Western Blot (2.5 µg/ml)
Alternate names:
Pro-ANP-converting enzyme; Low density lipoprotein receptor-related protein 4; Corin; Transmembrane protease serine 10; Heart-specific serine proteinase ATC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542916 309.00
Check your price
  • LRP1B Polyclonal Antibody for Western Blot
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547836
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Low-density lipoprotein receptor-related protein-deleted in tumor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547836 349.00
Check your price
  • LRP5 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 365400
  • Size 100µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Lr3; Low-density lipoprotein receptor-related protein 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
365400 309.00
Check your price
  • LRP8 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116913
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:500), Immunofluorescence (1:200-1:500), Western Blot (2 µg/ml)
Alternate names:
Apolipoprotein E receptor 2; Protein LR8B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116913 349.00
Check your price
  • LRP8 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Bovine, Chicken, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116914
  • Size 100µL
Applications
Immunocytochemistry (5 µg/ml), Immunofluorescence (5 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Apolipoprotein E receptor 2; Protein LR8B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116914 309.00
Check your price
  • LRP1 Monoclonal Antibody for Western Blot, IF, IHC, IHC (F) and Flow
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 373800
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (1:100), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Alpha-2-macroglobulin receptor; Apolipoprotein E receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
373800 305.00
Check your price
  • LRP8 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 407800
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Apolipoprotein E receptor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
407800 309.00
Check your price
  • LRP12 Isoform A Precursor Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521956
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Suppressor of tumorigenicity 7 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521956 349.00
Check your price
  • SORL1 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515369
  • Size 100µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Sorting protein-related receptor containing LDLR class A repeats; SorLA-1; Low-density lipoprotein receptor relative with 11 ligand-binding repeats
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515369 349.00
Check your price
  • SORL1 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA517941
  • Size 100ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Sorting protein-related receptor containing LDLR class A repeats; SorLA-1; Low-density lipoprotein receptor relative with 11 ligand-binding repeats; Gp250
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517941 349.00
Check your price
  • SORL1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Sheep
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547765
  • Size 100µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Sorting protein-related receptor containing LDLR class A repeats; SorLA-1; Low-density lipoprotein receptor relative with 11 ligand-binding repeats; Gp250
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547765 349.00
Check your price
  • SORL1 Monoclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA515355
  • Size 100µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Sorting protein-related receptor containing LDLR class A repeats; SorLA-1; Low-density lipoprotein receptor relative with 11 ligand-binding repeats
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515355 349.00
Check your price
  • SORL1 Polyclonal Antibody for IHC (F), IHC (P) and IHC (PFA)
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519482
  • Size 100µg
Applications
Immunohistochemistry (Frozen) (1:40), Immunohistochemistry (Paraffin) (1:40), Immunohistochemistry (PFA fixed) (1:40)
Alternate names:
Sorting protein-related receptor containing LDLR class A repeats; SorLA-1; Low-density lipoprotein receptor relative with 11 ligand-binding repeats
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519482 349.00
Check your price
  • LRP5 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518387
  • Size 100µg
Applications
Immunohistochemistry (3-6µg/ml)
Alternate names:
LRP5; low density lipoprotein receptor-related protein 5; HGNC:6697; BMND1; HBM; LR3; LRP7; OPPG; OPS; VBCH2; low density lipoprotein receptor-related protein 7; osteoporosis pseudoglioma syndrome; EVR1; EVR4; OPTA1; exudative vitreoretinopathy 1;
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518387 349.00
Check your price
  • RAP Polyclonal Antibody for Western Blot
  • Host Sheep
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547626
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Low density lipoprotein receptor-related protein-associated protein 1; Heparin-binding protein 44
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547626 349.00
Check your price
  • LRP5 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513144
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Lr3; Low-density lipoprotein receptor-related protein 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513144 349.00
Check your price
  • LRP6 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513145
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
LDL receptor-related protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513145 349.00
Check your price
  • LRP3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536486
  • Size 100µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
105 kDa low-density lipoprotein receptor-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536486 309.00
Check your price
  • LRP3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539230
  • Size 100µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
105 kDa low-density lipoprotein receptor-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539230 309.00
Check your price
  • LRP1 Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA127198
  • Size 100µg
Applications
Blocking Assay (Assay-dependent), Flow Cytometry (Assay Dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Paraffin, Frozen) (1-10 µg/ml), Western Blot (1-5 µg/ml)
Alternate names:
Apolipoprotein E receptor; Alpha-2-macroglobulin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA127198 375.00
Check your price
  • LRP12 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535069
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
Suppressor of tumorigenicity 7 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535069 349.00
Check your price
  • LRP1 Monoclonal Antibody for Western Blot, IF, IHC (F), Flow, IP and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180192
  • Size 200µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1/50-1/100), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay dependent (Non-redu)
Alternate names:
Apolipoprotein E receptor; Alpha-2-macroglobulin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180192 499.00
Check your price
  • LRP1 Monoclonal Antibody for Western Blot, IHC (F), Flow and ELISA
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 377600
  • Size 100µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Apolipoprotein E receptor; Alpha-2-macroglobulin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
377600 305.00
Check your price