1 - 30 of 30 products displayed
  • LOX-1 Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523895
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523895 275.00
Check your price
  • LOX-1 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547158
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547158 349.00
Check your price
  • LOX-1 Polyclonal Antibody for Western Blot, IHC (F), Flow and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA184920
  • Size 50 µg
Applications
Flow Cytometry (1:10), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Neat), Western Blot (1:10-1:100)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184920 399.00
Check your price
  • LOX-1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519804
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519804 349.00
Check your price
  • LOX-1 Monoclonal Antibody for Western Blot, Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524060
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524060 275.00
Check your price
  • LOX-1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532060
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532060 Not Orderable Online
Check your price
  • LOX-1 Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547205
  • Size 100 µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547205 349.00
Check your price
  • LOX-1 (soluble) Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516797
  • Size 200 ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516797 349.00
Check your price
  • ALOX15 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519052
  • Size 100 ug
Applications
Western Blot (0.2-0.5 ug/ml)
Alternate names:
Arachidonate omega-6 lipoxygenase; Arachidonate 12-lipoxygenase, leukocyte-type; 12/15-lipoxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519052 349.00
Check your price
  • LOX Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116953
  • Size 100 µL
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:100-1:250), Western Blot (0.5-4 µg/ml)
Alternate names:
Ras excision protein; Lysyl oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116953 375.00
Check your price
  • LOX Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA116955
  • Size 50 µL
Applications
Immunohistochemistry (Paraffin) (1:200), Western Blot (1:1000-1:5000)
Alternate names:
Ras excision protein; Lysyl oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116955 309.00
Check your price
  • LOX Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F) and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA146020
  • Size 100 µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100-1:250), Western Blot (1:4000)
Alternate names:
Ras excision protein; Lysyl oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146020 375.00
Check your price
  • ALOX15 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515065
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Arachidonate omega-6 lipoxygenase; Arachidonate 12-lipoxygenase, leukocyte-type; 12/15-lipoxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515065 349.00
Check your price
  • LOX Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526135
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Lysyl oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526135 349.00
Check your price
  • LOX Polyclonal Antibody for Western Blot, IF, ICC, IHC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA116989
  • Size 100 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:100-1:250), Western Blot (1:500-1:2000)
Alternate names:
Ras excision protein; Lysyl oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116989 Not Orderable Online
Check your price
  • LOX Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523001
  • Size 100 µL
Applications
Immunocytochemistry (1:10-1:500), Immunofluorescence (1:10-1:500), Immunohistochemistry (1:10-1:500), Western Blot (0.35 µg/ml)
Alternate names:
Ras excision protein; Lysyl oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523001 349.00
Check your price
  • LOX Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527270
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Lysyl oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527270 349.00
Check your price
  • ALOXE3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521833
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Hydroperoxy icosatetraenoate dehydratase; Epidermis-type lipoxygenase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521833 349.00
Check your price
  • LOX Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA523002
  • Size 100 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunoprecipitation (1:10-1:500), Western Blot (0.5 µg/ml)
Alternate names:
Lysyl oxidase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523002 349.00
Check your price
  • ALOX12 Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526020
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Platelet-type lipoxygenase 12; Lipoxin synthase 12-LO
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526020 349.00
Check your price
  • ALOX12B Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523608
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Epidermis-type lipoxygenase 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523608 349.00
Check your price
  • ALOX5 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA146002
  • Size 100 µL
Applications
Immunocytochemistry (1:500-1:2000), Immunofluorescence (1:500-1:2000), Western Blot (1:500-1:1000)
Alternate names:
5-lipoxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146002 309.00
Check your price
  • Phospho-ALOX5 (Ser271) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538408
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
5-LO, 5-LOX, 5LPG, LOG5, arachidonic 5-lipoxygenase alpha-10 isoform, arachidonic 5-lipoxygenase delta-10-13 isoform, arachidonic 5-lipoxygenase delta-13 isoform, arachidonic 5-lipoxygenase delta-p10 isoform, arachidonic acid 5-lipoxygenase, leukotriene A4 synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538408 309.00
Check your price
  • Phospho-ALOX5 (Ser523) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA539830
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
5-LO, 5-LOX, 5LPG, LOG5, arachidonic 5-lipoxygenase alpha-10 isoform, arachidonic 5-lipoxygenase delta-10-13 isoform, arachidonic 5-lipoxygenase delta-13 isoform, arachidonic 5-lipoxygenase delta-p10 isoform, arachidonic acid 5-lipoxygenase, leukotriene A4 synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539830 309.00
Check your price
  • ALOX15B Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526208
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Linoleate 13-lipoxygenase 15-LOb; Arachidonate 15-lipoxygenase type II; 15-lipoxygenase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526208 349.00
Check your price
  • ALOX5 Monoclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA514838
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:50), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
5-lipoxygenase; arachidonic acid 5-lipoxygenase; leukotriene A4 synthase
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514838 275.00
Check your price
  • ALOX5 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA518567
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
arachidonate 5-lipoxygenase; RP11-67C2.3; ALOX5; ALOX5; 5-LO; 5LPG; LOG5 ; arachidonic acid 5-lipoxygenase; leukotriene A4 synthase;
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518567 349.00
Check your price
  • ALOXE3 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523953
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Hydroperoxy icosatetraenoate dehydratase; Epidermis-type lipoxygenase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523953 349.00
Check your price
  • ALOX5 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA146003
  • Size 100 µL
Applications
Western Blot (1:500-1:1000)
Alternate names:
5-lipoxygenase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146003 309.00
Check your price
  • Phospho-ALOX5 (Ser523) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA14628
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
phospho-5-LO (Ser523)
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14628 349.00
Check your price