1 - 8 of 8 products displayed
  • LOX-1 Polyclonal Antibody for Western Blot, IHC (F), Flow and IP
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA184920
  • Size 50µg
Applications
Flow Cytometry (1:10), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Neat), Western Blot (1:10-1:100)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184920 399.00
Check your price
  • LOX-1 Monoclonal Antibody for Western Blot, Flow and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524060
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524060 275.00
Check your price
  • LOX-1 Polyclonal Antibody for Western Blot, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547205
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547205 349.00
Check your price
  • LOX-1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519804
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519804 349.00
Check your price
  • LOX-1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532060
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532060 349.00
Check your price
  • LOX-1 Monoclonal Antibody for Western Blot and Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523895
  • Size 100µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523895 275.00
Check your price
  • LOX-1 Polyclonal Antibody for Western Blot and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547158
  • Size 100µg
Applications
Blocking Assay (Assay-Dependent), Western Blot (0.1 µg/ml)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547158 349.00
Check your price
  • LOX-1 (soluble) Monoclonal Antibody for Western Blot and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516797
  • Size 200ug
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Lectin-type oxidized LDL receptor 1; Lectin-like oxidized LDL receptor 1; C-type lectin domain family 8 member A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516797 349.00
Check your price