1 - 2 of 2 products displayed
  • Cytokeratin 6/18 Antibody (Monoclonal, LP34)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134402
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Type-II keratin Kb6; Keratin-6A; Cytokeratin-6D; Cytokeratin-6A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134402 425.00
Check your price
  • Cytokeratin 6/18 Antibody (Monoclonal, LP34)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA134399
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Keratin-18; Cytokeratin-18; Cell proliferation-inducing gene 46 protein; Type-II keratin Kb6; Keratin-6A; Cytokeratin-6D; Cytokeratin-6A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA134399 375.00
Check your price