1 - 4 of 4 products displayed
  • EDG4 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513130
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Lysophosphatidic acid receptor Edg-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513130 349.00
Check your price
  • EDG4 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112694
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Lysophosphatidic acid receptor Edg-4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112694 550.00
Check your price
  • PPAP2A Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Guinea pig, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA542235
  • Size 100 µl
Applications
Immunohistochemistry (Assay-Dependent), Western Blot (5 µg/ml)
Alternate names:
Phosphatidic acid phosphatase 2a; Phosphatidate phosphohydrolase type 2a; PAP2-alpha; Lipid phosphate phosphohydrolase 1; Hydrogen peroxide-inducible protein 53; 35 kDa PAP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542235 309.00
Check your price
  • PPAP2A Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112678
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Phosphatidic acid phosphatase 2a; Phosphatidate phosphohydrolase type 2a; PAP2-alpha; Lipid phosphate phosphohydrolase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112678 309.00
Check your price