1 - 6 of 6 products displayed
  • EDG2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA11041
  • Size 100µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Lysophosphatidic acid receptor Edg-2; VZG-1; Rec1.3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA11041 349.00
Check your price
  • EDG2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA112306
  • Size 100µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Lysophosphatidic acid receptor Edg-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA112306 499.00
Check your price
  • EDG2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513128
  • Size 400µL
Applications
Western Blot (1:1000)
Alternate names:
Lysophosphatidic acid receptor Edg-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513128 349.00
Check your price
  • EDG2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528768
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Lysophosphatidic acid receptor Edg-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528768 349.00
Check your price
  • EDG2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533492
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml)
Alternate names:
Lysophosphatidic acid receptor Edg-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533492 349.00
Check your price
  • EDG2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533491
  • Size 50ug
Applications
Immunohistochemistry (Paraffin) (2-3 µg/ml)
Alternate names:
Lysophosphatidic acid receptor Edg-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533491 349.00
Check your price