1 - 11 of 11 products displayed
  • LRP6 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518317
  • Size 100 ug
Applications
Immunohistochemistry (4-6µg/ml)
Alternate names:
LRP6; low density lipoprotein receptor-related protein 6; low density lipoprotein-related protein 6; ADCAD2; FLJ90062; FLJ90421
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518317 349.00
Check your price
  • LRP6 Monoclonal Antibody for IF, ICC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523950
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml)
Alternate names:
LRP6, low density lipoprotein receptor-related protein 6, low density lipoprotein-related protein 6, ADCAD2, FLJ90062, FLJ90421
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523950 275.00
Check your price
  • LRP6 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547145
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Western Blot (0.1 µg/ml)
Alternate names:
LRP6, low density lipoprotein receptor-related protein 6, low density lipoprotein-related protein 6, ADCAD2, FLJ90062, FLJ90421
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547145 375.00
Check your price
  • LRP6 Monoclonal Antibody for Western Blot
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523995
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
LRP6, low density lipoprotein receptor-related protein 6, low density lipoprotein-related protein 6, ADCAD2, FLJ90062, FLJ90421
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523995 275.00
Check your price
  • LRP6 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513145
  • Size 400 µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
LDL receptor-related protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513145 349.00
Check your price
  • KREMEN1 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514549
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Kringle-containing protein marking the eye and the nose; Kringle domain-containing transmembrane protein 1; Dickkopf receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514549 349.00
Check your price
  • KREMEN1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA511624
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Kringle-containing protein marking the eye and the nose; Kringle domain-containing transmembrane protein 1; Dickkopf receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511624 349.00
Check your price
  • RSPO1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520749
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Roof plate-specific spondin-1; Cysteine-rich and single thrombospondin domain-containing protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520749 349.00
Check your price
  • LGR5 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA533728
  • Size 50 ug
Applications
Immunohistochemistry (Paraffin) (5 ug/ml)
Alternate names:
G-protein coupled receptor HG38; G-protein coupled receptor 67; G-protein coupled receptor 49
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA533728 349.00
Check your price
  • MESDC2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA535060
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (1:1000)
Alternate names:
Renal carcinoma antigen NY-REN-61; Mesoderm development protein; Mesoderm development candidate 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535060 349.00
Check your price
  • RSPO1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534937
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Roof plate-specific spondin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534937 349.00
Check your price