1 - 5 of 5 products displayed
  • LRRFIP2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Rat, Mouse, Human
  • Conjugate Unconjugated
  • Catalog # PA521115
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 µg/ml), Western Blot (0.5-1 µg/ml)
Alternate names:
HUFI-2; LRR FLII-interacting protein 2; leucine-rich repeat flightless-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521115 349.00
Check your price
  • LRRFIP2 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Horse, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523458
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
HUFI-2, LRR FLII-interacting protein 2, leucine-rich repeat flightless-interacting protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523458 349.00
Check your price
  • LRRFIP1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA523445
  • Size 100ug
Applications
Western Blot (2-5 µg/ml)
Alternate names:
H186 FLAP; FLI-LRR-associated protein 1; TAR RNA-interacting protein; GC-binding factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523445 349.00
Check your price
  • LRRFIP1 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521114
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 µg/ml), Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TAR RNA-interacting protein; GC-binding factor 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521114 349.00
Check your price
  • LRRFIP2 Synthetic Peptide for BLOCK and Ctrl
  • Host N/A
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PEP1229
  • Size 50ug
Applications
Blocking Assay (Assay Dependent), Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PEP1229 85.75
Check your price