1 - 2 of 2 products displayed
  • LRRK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513868
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513868 349.00
Check your price
  • LRRK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528565
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
RIPK6, Roco1, leucine-rich repeat serine/threonine-protein kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528565 349.00
Check your price