1 - 12 of 12 products displayed
  • LRRK2 Polyclonal Antibody for Western Blot, ICC, IHC, IHC (P), IM and pep-ELISA
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518319
  • Size 100ug
Applications
Immunocytochemistry (Assay Dependent), Immunohistochemistry (1-2µg/ml), Immunomicroscopy (Assay Dependent), Western Blot (0.3-1 ug/ml)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518319 349.00
Check your price
  • Phospho-LRRK2 (Ser935) Monoclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701066
  • Size 100µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (1:10-1:50), Western Blot (2-5 ug/ml)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
701066 304.00
Check your price
  • LRRK2 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511154
  • Size 400µL
Applications
Western Blot (1:100-1:500)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511154 349.00
Check your price
  • LRRK2 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511155
  • Size 400µL
Applications
Western Blot (1:100-1:500)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511155 349.00
Check your price
  • LRRK2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA146443
  • Size 100µg
Applications
Western Blot (1:500)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146443 309.00
Check your price
  • Phospho-LRRK2 (Ser935) Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710097
  • Size 100µg
Applications
Western Blot (2-5 ug/ml)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
710097 304.00
Check your price
  • LRRK2 Polyclonal Antibody for IHC (F) and IHC (P)
  • Host Sheep
  • Target Species Bovine, Human
  • Conjugate Unconjugated
  • Catalog # PA118254
  • Size 100µL
Applications
Immunohistochemistry (Paraffin, Frozen) (1:500)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA118254 349.00
Check your price
  • LRRK2 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (F), IHC (P) and Flow
  • Host Rabbit
  • Target Species Bovine, C. elegans, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA116770
  • Size 100µL
Applications
Immunocytochemistry (1:1000-1:2000), Immunofluorescence (1:1000-1:2000), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:500-1:1000), Immunoprecipitation (1:10-1:500), Western Blot (1:2000-1:5000)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116770 375.00
Check your price
  • LRRK2 Polyclonal Antibody for Western Blot, IF, ICC, IHC, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA146194
  • Size 100µL
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (1:10-1:500), Western Blot (1:1000)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146194 309.00
Check your price
  • LRRK2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513872
  • Size 400µL
Applications
Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513872 349.00
Check your price
  • LRRK2 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513874
  • Size 400µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513874 349.00
Check your price
  • LRRK2 Polyclonal Antibody for Western Blot, IHC and IHC (P)
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA116990
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (2.5 µg/ml), Western Blot (2 µg/ml)
Alternate names:
LRRK-2, LRRK 2, PARK8, ROCO2, Dardarin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116990 349.00
Check your price