1 - 2 of 2 products displayed
  • LSM11 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531328
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
2210404M20Rik, LSM11 homolog, U7 small nuclear RNA associated, U7 snRNA-associated Sm-like protein LSm11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531328 349.00
Check your price
  • LSM11 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531324
  • Size 100µL
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
2210404M20Rik, LSM11 homolog, U7 small nuclear RNA associated, U7 snRNA-associated Sm-like protein LSm11
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531324 349.00
Check your price