1 - 20 of 20 products displayed
  • Firefly luciferase Antibody (Monoclonal, Luci 21 1-107)
  • Host Mouse
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # MA116880
  • Size 100 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:1000)
Alternate names:
Luciferin 4-monooxygenase, Firefly Luciferase, Green Luciferase, Japanese firefly, Genji firefly, Photinus pyralis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116880 375.00
Check your price
  • Firefly luciferase Antibody (Monoclonal, Luci17)
  • Host Mouse
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # MA112556
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Luciferin 4-monooxygenase, Firefly Luciferase, Green Luciferase, Japanese firefly, Genji firefly, Photinus pyralis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112556 309.00
Check your price
  • Firefly luciferase Antibody (Monoclonal, CS 17)
  • Host Mouse
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # 356700
  • Size 100 µg
Applications
Immunofluorescence (10-30 ug/ml), Immunohistochemistry (Frozen) (15-50 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Luciferin 4-monooxygenase, Firefly Luciferase, Green Luciferase, Japanese firefly, Genji firefly, Photinus pyralis
Catalog # Size Price (AUD) Your Price (AUD) Qty
356700 304.00
Check your price
  • Firefly luciferase Antibody (Monoclonal, LCF01)
  • Host Mouse
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # MA190934
  • Size 500 µL
Applications
Immunoelectrophoresis (Assay-dependent), Western Blot (2 µg/ml)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190934 349.00
Check your price
  • Red Firefly Luciferase Antibody
  • Host Rabbit
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # PA1179
  • Size 100 µg
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunoprecipitation (5 µg), Neutralization (Assay dependent), Western Blot (1:2000-1:4000)
Alternate names:
Luciferin 4-monooxygenase, Firefly Luciferase, Red-shifted Firefly Luciferase, Japanese firefly, Genji firefly, Photinus pyralis
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1179 349.00
Check your price
  • Cypridina luciferase Antibody
  • Host Rabbit
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # PA1182
  • Size 100 ug
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
Cypridina luciferase, Cypridina noctiluca luciferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1182 309.00
Check your price
  • Gaussia luciferase Antibody
  • Host Rabbit
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # PA1181
  • Size 100 ug
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
Gaussia luciferase, Gaussia princeps luciferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1181 309.00
Check your price
  • TurboLuc luciferase Antibody
  • Host Rabbit
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # PA1178
  • Size 100 ug
Applications
Western Blot (1:1000)
Alternate names:
TurboLuc luciferase synthetic
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1178 309.00
Check your price
  • Firefly luciferase Antibody (Monoclonal, LUC-Y)
  • Host Mouse
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # MA180225
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (10 ug/ml), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1/100)
Alternate names:
Luciferin 4-monooxygenase, Firefly Luciferase, Green Luciferase, Japanese firefly, Genji firefly, Photinus pyralis
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180225 449.00
Check your price
  • Firefly luciferase Antibody
  • Host Chicken
  • Target Species Luciferase
  • Conjugate Unconjugated
  • Catalog # PA127518
  • Size 50 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
Luciferin 4-monooxygenase, Firefly Luciferase, Green Luciferase, Japanese firefly, Genji firefly, Photinus pyralis
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127518 449.00
Check your price
  • Green Renilla luciferase Antibody
  • Host Rabbit
  • Target Species Renilla
  • Conjugate Unconjugated
  • Catalog # PA1180
  • Size 100 µg
Applications
Immunocytochemistry (1:500-1:1000), Immunofluorescence (1:500-1:1000), Immunoprecipitation (5 µg), Neutralization (Assay Dependent), Western Blot (1:2000-1:4000)
Alternate names:
Renilla-luciferin 2-monooxygenase, EC 1.13.12.5, Renilla-type luciferase, Green-shifted Renilla luciferase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1180 349.00
Check your price
  • Transferrin Receptor Antibody (Monoclonal, H68.4)
  • Host Mouse
  • Target Species Chicken, Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 136800
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Immunomicroscopy (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
CD71, T9, TFR, TFR1, TR, TRFR, p90, transferrin receptor protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
136800 312.00
Check your price
  • SMAD3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 511500
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Flow Cytometry (1:100-1:500), Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (1.0 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
SMAD family member 3; JV15-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
511500 305.00
Check your price
  • ZBED3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520767
  • Size 100 ug
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Axin-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520767 349.00
Check your price
  • RWDD3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520821
  • Size 100 ug
Applications
Western Blot (0.5 µg/ml)
Alternate names:
RWD domain-containing sumoylation enhancer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520821 349.00
Check your price
  • RWDD3 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA520820
  • Size 100 ug
Applications
Western Blot (1 µg/ml)
Alternate names:
RWD domain-containing sumoylation enhancer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520820 349.00
Check your price
  • ZBED3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA520733
  • Size 100 ug
Applications
Immunocytochemistry (20 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Axin-interacting protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520733 349.00
Check your price
  • SMAD2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 511300
  • Size 100 µg
Applications
ChIP assay (1-3 µg x 10^6 cells), ELISA (0.1-1.0 ug/ml), Flow Cytometry (3-5 µg x 10^6 cells), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1-3 ug/ml)
Alternate names:
SMAD family member 2; Mad-related protein 2; JV18-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
511300 305.00
Check your price
  • RWDD3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534884
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
RWD domain-containing sumoylation enhancer
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534884 349.00
Check your price
  • DnaJ Antibody
  • Host Mouse
  • Target Species Bacteria
  • Conjugate Unconjugated
  • Catalog # PA3018
  • Size 100 µL
Applications
Western Blot (1:100)
Alternate names:
Escherichia coli, DNAJA2, CPR3, DNJ3 , HDJ-2,
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3018 349.00
Check your price