1 - 6 of 6 products displayed
  • IKAROS Monoclonal Antibody for IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516418
  • Size 500µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Lymphoid transcription factor LyF-1; Ikaros family zinc finger protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516418 499.00
Check your price
  • IKAROS Polyclonal Antibody for Western Blot, IF, ICC, Flow and ChIP
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547682
  • Size 100µg
Applications
ChIP assay (5 µg/5 x 10^6 cells), Flow Cytometry (2.5 µg/10^6 cells), Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Western Blot (1-10 µg/ml)
Alternate names:
Lymphoid transcription factor LyF-1; Ikaros family zinc finger protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547682 375.00
Check your price
  • IKAROS Polyclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523728
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Lymphoid transcription factor LyF-1; Ikaros family zinc finger protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523728 349.00
Check your price
  • IKAROS Polyclonal Antibody for Western Blot and IHC (P)
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519214
  • Size 100ug
Applications
Immunohistochemistry (Paraffin) (2-4ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
Lymphoid transcription factor LyF-1; Ikaros family zinc finger protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519214 349.00
Check your price
  • IKAROS Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532451
  • Size 500µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Lymphoid transcription factor LyF-1; Ikaros family zinc finger protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532451 349.00
Check your price
  • IKAROS Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518735
  • Size 100ug
Applications
Western Blot (0.01-0.03 ug/ml)
Alternate names:
Lymphoid transcription factor LyF-1; Ikaros family zinc finger protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518735 349.00
Check your price