1 - 30 of 30 products displayed
  • Phospho-MEK1 + MEK2 pSer217 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Mouse, Rat, Human
  • Conjugate
  • Catalog # PA535603
  • Size 100µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535603 339.00
Check your price
  • Phospho-MEK1/MEK2 (Ser221) Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA126646
  • Size 50 µL
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Western Blot (0.1-1 µg/ml)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA126646 349.00
Check your price
  • Phospho-MEK1/MEK2 (Ser218, Ser222) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536823
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536823 349.00
Check your price
  • Phospho-MEK1/MEK2 (Ser222, Ser226) Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536824
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536824 349.00
Check your price
  • MEK1/MEK2 Monoclonal Antibody for Western Blot, IF, ICC, IHC (P) and Flow
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515135
  • Size 100 µL
Applications
Flow Cytometry (1:25), Immunocytochemistry (1:25), Immunofluorescence (1:25), Immunohistochemistry (Paraffin) (1:25), Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515135 275.00
Check your price
  • Phospho-MEK1/MEK2 (Ser218, Ser222, Ser226) Polyclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44454G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Western Blot (1:500-1:2000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44454G 309.00
Check your price
  • Phospho-MEK1/MEK2 (Ser222) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44452
  • Size 10 blots
Applications
Western Blot (1:1,000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44452 309.00
Check your price
  • Phospho-MEK1/MEK2 (Ser217, Ser221) Monoclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Chicken, Fruit fly, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515016
  • Size 100 µL
Applications
Immunoprecipitation (1:100), Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1; Protein rolled; Extracellular-regulated kinase A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515016 309.00
Check your price
  • Phospho-MEK1/MEK2 (Ser221) Monoclonal Antibody for Western Blot, IHC (P) and Flow
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515118
  • Size 100 µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:50), Western Blot (1:2000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515118 309.00
Check your price
  • MEK1/MEK2 Monoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515162
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515162 275.00
Check your price
  • MEK1/MEK2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531918
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1; MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531918 349.00
Check your price
  • MEK1/MEK2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA531917
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531917 349.00
Check your price
  • MEK1/MEK2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA110034
  • Size 50 ug
Applications
Western Blot (0.5 ug/ml)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1; MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA110034 159.00
Check your price
  • MEK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515161
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515161 349.00
Check your price
  • Phospho-MEK1/MEK2 (Ser217, Ser221) Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA14606
  • Size 100 µL
Applications
Western Blot (1:1,000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2; MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA14606 349.00
Check your price
  • Phospho-MEK2 (Thr394) Polyclonal Antibody for Western Blot, IHC (P) and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538140
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538140 309.00
Check your price
  • MEK1 Monoclonal Antibody for Western Blot and IP
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515093
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:2000)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515093 275.00
Check your price
  • MEK1 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517677
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517677 275.00
Check your price
  • MEK2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517678
  • Size 100 µL
Applications
Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517678 275.00
Check your price
  • MEK2 Monoclonal Antibody for Western Blot, IF, ICC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 700829
  • Size 100 µg
Applications
Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (5 ug/ml), Immunofluorescence (5 ug/ml), Western Blot (0.5-1µg/ml)
Alternate names:
MAPK/ERK kinase 2; ERK activator kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
700829 309.00
Check your price
  • Phospho-MEK1 (Thr286) Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA517664
  • Size 100 µL
Applications
Flow Cytometry (1:50), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 1; ERK activator kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517664 309.00
Check your price
  • Phospho-c-Raf (Tyr340, Tyr341) Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44506G
  • Size 10 blots
Applications
Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Raf-1; Proto-oncogene c-RAF
Catalog # Size Price (AUD) Your Price (AUD) Qty
44506G Not Orderable Online
Check your price
  • MEKK1 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA515085
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA515085 349.00
Check your price
  • ERK1 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137638
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137638 349.00
Check your price
  • MEK3 Monoclonal Antibody for Western Blot
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517216
  • Size 100 µL
Applications
Western Blot (2 ug/ml)
Alternate names:
MAPK/ERK kinase 3; Stress-activated protein kinase kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517216 349.00
Check your price
  • MEK3 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA517480
  • Size 100 µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
MAPK/ERK kinase 3; Stress-activated protein kinase kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517480 275.00
Check your price
  • ERK5 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514416
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Extracellular signal-regulated kinase 5; Big MAP kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514416 349.00
Check your price
  • Phospho-c-Raf (Ser471) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512767
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
Raf-1; Proto-oncogene c-RAF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512767 349.00
Check your price
  • Phospho-c-Raf (Tyr340) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512875
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
Raf-1; Proto-oncogene c-RAF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512875 349.00
Check your price
  • Phospho-c-Raf (Ser494) Polyclonal Antibody for DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512890
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
Raf-1; Proto-oncogene c-RAF
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512890 349.00
Check your price