1 - 4 of 4 products displayed
  • MTNR1B Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532934
  • Size 50 ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (5 µg/ml)
Alternate names:
FGQTL2, MEL-1B-R, MT2, mel1b receptor, melatonin receptor MEL1B, melatonin receptor type 1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532934 349.00
Check your price
  • MTNR1B Polyclonal Antibody for IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532935
  • Size 50 ug
Applications
ELISA (Assay-dependent), Immunohistochemistry (Paraffin) (10 µg/ml)
Alternate names:
FGQTL2, MEL-1B-R, MT2, mel1b receptor, melatonin receptor MEL1B, melatonin receptor type 1B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532935 349.00
Check your price
  • MTNR1B Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132563
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
MT2, MTNR1b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132563 399.00
Check your price
  • MTNR1B Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA132564
  • Size 25 µg
Applications
Immunohistochemistry (Paraffin) (1:100-1:200)
Alternate names:
MT2, MTNR1b
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132564 399.00
Check your price