1 - 30 of 30 products displayed
  • TGF beta-2 Oligoclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 710276
  • Size 100µg
Applications
Western Blot (1-2 ug/ml)
Alternate names:
Polyergin; Glioblastoma-derived T-cell suppressor factor; Cetermin; BSC-1 cell growth inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
710276 309.00
Check your price
  • PKD2 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529834
  • Size 100µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Transient receptor potential cation channel subfamily P member 2; R48321; Polycystwin; Polycystic kidney disease 2 protein; Autosomal dominant polycystic kidney disease type II protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529834 349.00
Check your price
  • IGF2BP2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529870
  • Size 100µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
VICKZ family member 2; IGF-II mRNA-binding protein 2; Hepatocellular carcinoma autoantigen p62
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529870 349.00
Check your price
  • Bcl-2 Monoclonal Antibody for Western Blot, IF, IHC, Flow and IP
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 138800
  • Size 100µg
Applications
Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
PPP1R50, apoptosis regulator Bcl-2, protein phosphatase 1 regulatory subunit 50
Catalog # Size Price (AUD) Your Price (AUD) Qty
138800 305.00
Check your price
  • CXCL2 Monoclonal Antibody for Western Blot, ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523737
  • Size 500µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (1-4 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Macrophage inflammatory protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523737 375.00
Check your price
  • PKD2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSP00018W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Transient receptor potential cation channel subfamily P member 2; R48321; Polycystwin; Polycystic kidney disease 2 protein; Autosomal dominant polycystic kidney disease type II protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSP00018W 309.00
Check your price
  • AP2 alpha/beta Monoclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190265
  • Size 100µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Frozen) (Assay dependent), Immunohistochemistry (Paraffin) (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Activating enhancer-binding protein 2-beta; Activator protein 2; Activating enhancer-binding protein 2-alpha; AP-2 transcription factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190265 425.00
Check your price
  • ONECUT2 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524275
  • Size 100µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Transcription factor ONECUT-2; One cut homeobox 2; Hepatocyte nuclear factor 6-beta
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524275 275.00
Check your price
  • CD86 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517953
  • Size 50ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
Early T-cell costimulatory molecule 1; Activation B7-2 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517953 349.00
Check your price
  • CD86 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA517954
  • Size 300ug
Applications
Flow Cytometry (1.0 ug/10^6 cells)
Alternate names:
Early T-cell costimulatory molecule 1; Activation B7-2 antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517954 1,700.00
Check your price
  • CUG-BP Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536277
  • Size 100µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
RNA-binding protein BRUNOL-3; Neuroblastoma apoptosis-related RNA-binding protein; ELAV-type RNA-binding protein 3; CUG-BP- and ETR-3-like factor 2; CUG triplet repeat RNA-binding protein 2; Bruno-like protein 3; RNA-binding protein BRUNOL-2; Embryo deadenylation element-binding protein homolog; Deadenylation factor CUG-BP; CUG-BP- and ETR-3-like factor 1; CUG triplet repeat RNA-binding protein 1; Bruno-like protein 2; 50 kDa nuclear polyadenylated RNA-binding protein; Deadenylation factor EDEN-BP; Brain protein F41; SAPK gamma
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536277 309.00
Check your price
  • SERPINF2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA522085
  • Size 100µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Serpin F2; Alpha-2-plasmin inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522085 349.00
Check your price
  • CXCL2 Polyclonal Antibody for Western Blot, IF, ICC, IHC and Neu
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547015
  • Size 100µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (5-15 µg/ml), Neutralization (0.015-0.075 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Macrophage inflammatory protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547015 375.00
Check your price
  • KCNA2 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Pig, Bovine, Dog, Human, Mouse, Rat, Xenopus, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA523005
  • Size 100µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500-1:1000)
Alternate names:
Voltage-gated potassium channel subunit Kv1.2; Voltage-gated potassium channel HBK5; Voltage-gated K(+) channel HuKIV; NGK1; CSMK1; RCK5; RBK2; RAK; MK2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523005 349.00
Check your price
  • ERLIN2 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA517103
  • Size 100µL
Applications
Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Stomatin-prohibitin-flotillin-HflC/K domain-containing protein 2; Endoplasmic reticulum lipid raft-associated protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA517103 275.00
Check your price
  • CXCL2 Polyclonal Antibody for Western Blot and IP
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA128895
  • Size 100µg
Applications
Immunoprecipitation (1:400-1:600), Western Blot (1:1500-1:2500)
Alternate names:
Macrophage inflammatory protein 2; Cytokine-induced neutrophil chemoattractant 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128895 309.00
Check your price
  • SERPINF2 Polyclonal Antibody for Western Blot, IP and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547142
  • Size 100µg
Applications
Immunoprecipitation (25 µg/ml), Neutralization (4.2 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Serpin F2; Alpha-2-plasmin inhibitor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547142 349.00
Check your price
  • CXCL2 Polyclonal Antibody for ELISA
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # ARC1074
  • Size 500µg
Applications
ELISA (Assay Dependent)
Alternate names:
Macrophage inflammatory protein 2; Cytokine-induced neutrophil chemoattractant 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
ARC1074 309.00
Check your price
  • CXCL2 Polyclonal Antibody, Biotin conjugate for Western Blot and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Biotin
  • Catalog # PA132940
  • Size 50µg
Applications
ELISA (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Macrophage inflammatory protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132940 650.00
Check your price
  • NKCC1 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Bovine, Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate
  • Catalog # PA127915
  • Size 50µg
Applications
Western Blot (1:2000)
Alternate names:
Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1; Basolateral Na-K-Cl symporter
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127915 Not Orderable Online
Check your price
  • TAS1R2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # OSR00183W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Sweet taste receptor T1R2; G-protein coupled receptor 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00183W 309.00
Check your price
  • TAS1R2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # OSR00180W
  • Size 100µL
Applications
Immunohistochemistry (1:300-1:2000), Western Blot (1:300-1:2000)
Alternate names:
Sweet taste receptor T1R2; G-protein coupled receptor 71
Catalog # Size Price (AUD) Your Price (AUD) Qty
OSR00180W 309.00
Check your price
  • GREM1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA513123
  • Size 400µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Increased in high glucose protein 2; Down-regulated in Mos-transformed cells protein; DAN domain family member 2; Cysteine knot superfamily 1, BMP antagonist 1; Cell proliferation-inducing gene 2 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513123 349.00
Check your price
  • ESRRB Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1315
  • Size 100µg
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Western Blot (2 ug/ml)
Alternate names:
Nuclear receptor subfamily 3 group B member 2; Estrogen-related receptor beta; Estrogen receptor-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1315 375.00
Check your price
  • LIMP2 Polyclonal Antibody for Western Blot and IHC
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520544
  • Size 100ug
Applications
Immunohistochemistry (10 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Scavenger receptor class B member 2; Lysosome membrane protein II; 85 kDa lysosomal membrane sialoglycoprotein; CD36 antigen-like 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520544 349.00
Check your price
  • IGF2BP2 Polyclonal Antibody for Western Blot, IHC and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA523636
  • Size 400µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
VICKZ family member 2; IGF-II mRNA-binding protein 2; Hepatocellular carcinoma autoantigen p62
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523636 349.00
Check your price
  • MCP-2 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701174
  • Size 100µg
Applications
Western Blot (2-3 ug/ml)
Alternate names:
Small-inducible cytokine A8; Monocyte chemotactic protein 2; Monocyte chemoattractant protein 2; HC14
Catalog # Size Price (AUD) Your Price (AUD) Qty
701174 309.00
Check your price
  • Phospho-SMAD2 (Ser118) Polyclonal Antibody for IF, ICC and DB
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513083
  • Size 400µL
Applications
Dot blot (1:500), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50)
Alternate names:
SMAD family member 2; Mad-related protein 2; JV18-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513083 349.00
Check your price
  • SLC39A2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA542462
  • Size 100µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Zrt- and Irt-like protein 2; Solute carrier family 39 member 2; Eti-1; 6A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542462 309.00
Check your price
  • SLC39A2 Polyclonal Antibody for Western Blot, IF, ICC and IHC
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA521067
  • Size 100ug
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Zrt- and Irt-like protein 2; Solute carrier family 39 member 2; Eti-1; 6A1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521067 349.00
Check your price