1 - 30 of 30 products displayed
  • SLC45A2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542416
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Solute carrier family 45 member 2; Protein underwhite; Melanoma antigen AIM1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542416 309.00
Check your price
  • PITPNM3 Polyclonal Antibody for IHC
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518269
  • Size 100 ug
Applications
Immunohistochemistry (5-10µg/ml)
Alternate names:
Pyk2 N-terminal domain-interacting receptor 1; Phosphatidylinositol transfer protein, membrane-associated 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518269 349.00
Check your price
  • PITPNM1 Polyclonal Antibody for Western Blot and ICC
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA518266
  • Size 100 ug
Applications
Immunocytochemistry (Assay Dependent), Western Blot (2 ug/ml)
Alternate names:
Pyk2 N-terminal domain-interacting receptor 2; Phosphatidylinositol transfer protein, membrane-associated 1; Mpt-1; Drosophila retinal degeneration B homolog 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518266 349.00
Check your price
  • PMCA4 ATPase Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow and ELISA
  • Host Mouse
  • Target Species Bovine, Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA1914
  • Size 100 µg
Applications
ELISA (1:15,000), Flow Cytometry (2µg per 10^6 cells), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (1:100-1:500), Western Blot (1:1,000)
Alternate names:
Plasma membrane calcium pump isoform 4; Plasma membrane calcium ATPase isoform 4; Matrix-remodeling-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1914 375.00
Check your price
  • PMCA ATPase Monoclonal Antibody for Western Blot, IF, ICC, IHC (F), IHC (P), Flow and IP
  • Host Mouse
  • Target Species Amphibian, Dog, Chicken, Cat, Human, Mouse, Non-human primate, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA3914
  • Size 100 µL
Applications
Flow Cytometry (1ug/test), Immunocytochemistry (Assay dependent), Immunofluorescence (1:100-1:200), Immunohistochemistry (Paraffin, Frozen) (1:500), Immunoprecipitation (Assay dependent), Western Blot (1:1000-1:5000)
Alternate names:
Plasma membrane calcium pump isoform 1; Plasma membrane calcium ATPase isoform 1; Plasma membrane calcium pump isoform 2; Plasma membrane calcium ATPase isoform 2; Plasma membrane calcium pump isoform 3; Plasma membrane calcium ATPase isoform 3; Plasma membrane calcium pump isoform 4; Plasma membrane calcium ATPase isoform 4; Matrix-remodeling-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA3914 375.00
Check your price
  • PITPNM3 Polyclonal Antibody for Western Blot, IF and ICC
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521903
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Pyk2 N-terminal domain-interacting receptor 1; Phosphatidylinositol transfer protein, membrane-associated 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521903 349.00
Check your price
  • PMCA3 ATPase Polyclonal Antibody for Western Blot, IHC (F), IHC (P) and ELISA
  • Host Rabbit
  • Target Species Human, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA1916
  • Size 100 µg
Applications
ELISA (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:20), Western Blot (1:1,000)
Alternate names:
Plasma membrane calcium pump isoform 3; Plasma membrane calcium ATPase isoform 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1916 349.00
Check your price
  • GOT2 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515595
  • Size 100 µL
Applications
Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Transaminase A; Plasma membrane-associated fatty acid-binding protein; Kynurenine--oxoglutarate transaminase IV; Kynurenine--oxoglutarate transaminase 4; Kynurenine aminotransferase IV; Kynurenine aminotransferase 4; Glutamate oxaloacetate transaminase 2; Fatty acid-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515595 349.00
Check your price
  • GOT2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA527572
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Transaminase A; Plasma membrane-associated fatty acid-binding protein; Kynurenine--oxoglutarate transaminase IV; Kynurenine--oxoglutarate transaminase 4; Kynurenine aminotransferase IV; Kynurenine aminotransferase 4; Glutamate oxaloacetate transaminase 2; Fatty acid-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527572 349.00
Check your price
  • GOT2 Polyclonal Antibody for Western Blot and Flow
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535374
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Transaminase A; Plasma membrane-associated fatty acid-binding protein; Kynurenine--oxoglutarate transaminase IV; Kynurenine--oxoglutarate transaminase 4; Kynurenine aminotransferase IV; Kynurenine aminotransferase 4; Glutamate oxaloacetate transaminase 2; Fatty acid-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535374 349.00
Check your price
  • GOT2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518835
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
Transaminase A; Plasma membrane-associated fatty acid-binding protein; Kynurenine--oxoglutarate transaminase IV; Kynurenine--oxoglutarate transaminase 4; Kynurenine aminotransferase IV; Kynurenine aminotransferase 4; Glutamate oxaloacetate transaminase 2; Fatty acid-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518835 349.00
Check your price
  • GOT2 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518869
  • Size 100 ug
Applications
Western Blot (0.03-0.1 ug/ml)
Alternate names:
Transaminase A; Plasma membrane-associated fatty acid-binding protein; Kynurenine--oxoglutarate transaminase IV; Kynurenine--oxoglutarate transaminase 4; Kynurenine aminotransferase IV; Kynurenine aminotransferase 4; Glutamate oxaloacetate transaminase 2; Fatty acid-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518869 349.00
Check your price
  • MARCH3 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA539145
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Membrane-associated RING-CH protein III; Membrane-associated RING finger protein 3; RING finger protein 173
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539145 309.00
Check your price
  • MARCH2 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530620
  • Size 100 µL
Applications
Western Blot (1:1000-1:10,000)
Alternate names:
RING finger protein 172; Membrane-associated RING-CH protein II; Membrane-associated RING finger protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530620 349.00
Check your price
  • PMCA3 ATPase Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530936
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Plasma membrane calcium pump isoform 3; Plasma membrane calcium ATPase isoform 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530936 349.00
Check your price
  • CDH17 Polyclonal Antibody for Western Blot, IF and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA536295
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Liver-intestine cadherin; Intestinal peptide-associated transporter HPT-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536295 309.00
Check your price
  • CDH17 Polyclonal Antibody for Western Blot
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127949
  • Size 50µg
Applications
Western Blot (1:2000)
Alternate names:
Liver-intestine cadherin; Intestinal peptide-associated transporter HPT-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127949 425.00
Check your price
  • LAX1 Polyclonal Antibody for Western Blot and Flow
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547652
  • Size 100 µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Western Blot (1 µg/ml)
Alternate names:
Membrane-associated adapter protein LAX; Linker for activation of X cells
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547652 349.00
Check your price
  • MAGI2 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA547901
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Membrane-associated guanylate kinase inverted 2; Atrophin-1-interacting protein 1; Activin receptor-interacting protein 1; Atrophin-1-interacting protein A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547901 349.00
Check your price
  • MARCH8 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542273
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
RING finger protein 178; Membrane-associated RING-CH protein VIII; Membrane-associated RING finger protein 8; Cellular modulator of immune recognition
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542273 309.00
Check your price
  • MARCH8 Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Bovine, Dog, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA542272
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
RING finger protein 178; Membrane-associated RING-CH protein VIII; Membrane-associated RING finger protein 8; Cellular modulator of immune recognition
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542272 309.00
Check your price
  • MARCH10 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA518853
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
RING finger protein 190; Membrane-associated RING-CH protein X; Membrane-associated RING finger protein 10
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518853 349.00
Check your price
  • LAX1 Polyclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530891
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Membrane-associated adapter protein LAX; Linker for activation of X cells
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530891 349.00
Check your price
  • GOLGA3 Polyclonal Antibody for Western Blot
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518419
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
Golgin-160; Golgi complex-associated protein of 170 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518419 349.00
Check your price
  • LAP (Latency Associated Peptide) Monoclonal Antibody for Western Blot, IHC and Flow
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517186
  • Size 100 ug
Applications
Flow Cytometry (1:200-1:400), Immunohistochemistry (1:200-1:1000), Western Blot (1:500-1:2000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517186 309.00
Check your price
  • LAP (Latency Associated Peptide) Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523701
  • Size 500 µg
Applications
ELISA (0.5-2 µg/ml (Detection), 2-8 µg/ml (Capture))
Alternate names:
TGFB, TGFb-1, DPD1, Transforming Growth Factor, Transforming Growth Factor Beta-1, TGF-beta-1, CED
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523701 349.00
Check your price
  • GOLGA3 Polyclonal Antibody for Western Blot, IF, ICC and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA521299
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Golgin-160; Golgi complex-associated protein of 170 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521299 349.00
Check your price
  • Podoplanin Monoclonal Antibody for IHC (F) and IHC (P)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182747
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (0.75-2.0 µg/ml), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
T1-alpha; PA2.26 antigen; Glycoprotein 36; Aggrus
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182747 425.00
Check your price
  • Podoplanin Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537285
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
T1-alpha; PA2.26 antigen; Glycoprotein 36; Aggrus; Transmembrane glycoprotein E11; OTS-8; Glycoprotein 38; Type I cell 40 kDa protein; RTI140; E11 antigen epitope
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537285 309.00
Check your price
  • Furin Polyclonal Antibody for Western Blot, IF, ICC, Flow and IP
  • Host Rabbit
  • Target Species Dog, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1062
  • Size 100 µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunoprecipitation (1:100), Western Blot (1:1,000)
Alternate names:
Paired basic amino acid residue-cleaving enzyme; Dibasic-processing enzyme; Prohormone convertase 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1062 375.00
Check your price