1 - 2 of 2 products displayed
  • MBD3L2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA545867
  • Size 100 µl
Applications
Western Blot (1 µg/ml )
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA545867 309.00
Check your price
  • MBD1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1889
  • Size 100 µg
Applications
Western Blot (2-5 µg/ml)
Alternate names:
Protein containing methyl-CpG-binding domain 1; Methyl-CpG-binding protein MBD1; CXXC-type zinc finger protein 3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1889 349.00
Check your price