1 - 30 of 30 products displayed
  • HLA Class I Antibody, FITC (Monoclonal, 2G5)
  • Host Mouse
  • Target Species Bovine, Guinea pig, Hamster, Human, Mouse, Sheep, Pig, Rat
  • Conjugate FITC
  • Catalog # MA181300
  • Size 100 µg
Applications
Flow Cytometry (Neat-1:5)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181300 349.00
Check your price
  • HLA Class I Antibody (Monoclonal, 2G5)
  • Host Mouse
  • Target Species Bovine, Guinea pig, Hamster, Human, Mouse, Sheep, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA180907
  • Size 250 µg
Applications
Flow Cytometry (1/10-1/25), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180907 425.00
Check your price
  • HLA-E Antibody (Monoclonal, MEM-E/02)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119309
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-10 µg/ml), Western Blot (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119309 349.00
Check your price
  • HLA-ABC Antibody (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA511723
  • Size 500 µL
Applications
Flow Cytometry (0.5 ug/test), Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1:10-1:200)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511723 309.00
Check your price
  • HLA-ABC Antibody (Monoclonal, YTH862.2)
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA180014
  • Size 200 µg
Applications
Flow Cytometry (1/50-1/100), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180014 399.00
Check your price
  • HLA-ABC Antibody (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA170111
  • Size 500 ug
Applications
Flow Cytometry (1 ug/1 million cells), Functional Assay (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170111 309.00
Check your price
  • HLA Class I Antibody, FITC (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # AHU0228
  • Size 100 tests
Applications
Flow Cytometry (1:300)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHU0228 459.00
Check your price
  • HLA-ABC Antibody, Biotin (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate, Shrew
  • Conjugate Biotin
  • Catalog # MA180453
  • Size 100 µg
Applications
Flow Cytometry (Neat-1/10)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180453 349.00
Check your price
  • HLA-ABC Antibody, FITC (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate, Shrew
  • Conjugate FITC
  • Catalog # MA180454
  • Size 100 µg
Applications
Flow Cytometry (Neat)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180454 309.00
Check your price
  • HLA-ABC Antibody, PE (Monoclonal, MEM-123)
  • Host Mouse
  • Target Species Non-human primate, Bovine, Human
  • Conjugate PE
  • Catalog # MA110346
  • Size 100 ug
Applications
Flow Cytometry (1-2 ug/ml)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110346 325.00
Check your price
  • HLA-ABC Antibody, PE (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate, Shrew
  • Conjugate PE
  • Catalog # MA180455
  • Size 100 Tests
Applications
Flow Cytometry (Neat)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180455 349.00
Check your price
  • HLA-ABC Antibody, PE (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Cat, Human, Non-human primate
  • Conjugate PE
  • Catalog # MA119662
  • Size 100 µg
Applications
Flow Cytometry (1:50)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119662 375.00
Check your price
  • HLA-ABC Antibody (Monoclonal, MEM-123)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119312
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119312 225.00
Check your price
  • HLA-ABC Antibody (Monoclonal, MEM-147)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119151
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119151 225.00
Check your price
  • HLA-ABC Antibody (Monoclonal, MEM-81)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119153
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119153 225.00
Check your price
  • HLA-ABC Antibody (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Human, Non-human primate, Rodent
  • Conjugate Unconjugated
  • Catalog # MA180123
  • Size 500 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (1:50-1:100), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA180123 550.00
Check your price
  • HLA-ABC Antibody (Monoclonal, W6/32)
  • Host Mouse
  • Target Species Bovine, Cat, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119027
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Functional Assay (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119027 349.00
Check your price
  • HLA-ABC Antibody (Monoclonal, ICO-53)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA17660
  • Size 100 ug
Applications
Flow Cytometry (Assay dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17660 299.00
Check your price
  • HLA-ABC Antibody (Monoclonal, YTH 862.2)
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA190468
  • Size 100 µg
Applications
Flow Cytometry (1:50-1:100 (10 µl for 10^6 cells in 100 µl)), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen B*8; MHC class I antigen A*3; MHC class I antigen B*7; MHC class I antigen Cw*7
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA190468 449.00
Check your price
  • HLA-B7 Antibody (Monoclonal, BB7.1)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA182002
  • Size 200 µg
Applications
Flow Cytometry (1/50)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182002 399.00
Check your price
  • HLA-B7 Antibody, FITC (Monoclonal, BB7.1)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate FITC
  • Catalog # MA182180
  • Size 100 µg
Applications
Flow Cytometry (Neat-1/10)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182180 375.00
Check your price
  • Clathrin Heavy Chain Antibody (Monoclonal, X22)
  • Host Mouse
  • Target Species Bovine, Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1065
  • Size 100 µL
Applications
Flow Cytometry (1/200), Immunocytochemistry (Assay dependent), Immunofluorescence (1:1,000), Immunohistochemistry (1:100), Immunomicroscopy (Assay Dependent), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:500)
Alternate names:
Clathrin heavy chain on chromosome 17
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1065 375.00
Check your price
  • HLA-G Antibody, Biotin (Monoclonal, MEM-G/1)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA119512
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119512 309.00
Check your price
  • HLA-G Antibody (Monoclonal, MEM-G/2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119394
  • Size 100 µg
Applications
Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119394 249.00
Check your price
  • HLA-G Antibody (Monoclonal, MEM-G/1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119219
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin, Frozen) (1:60-1:100), Western Blot (Assay Dependent)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119219 249.00
Check your price
  • HLA-G Antibody (Monoclonal, MEM-G/4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119358
  • Size 100 µg
Applications
Western Blot (Assay Dependent)
Alternate names:
MHC class I antigen G; HLA G antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119358 249.00
Check your price
  • HLA-ABC Antibody (Monoclonal, 246-B8.E7)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA513582
  • Size 500 µL
Applications
Flow Cytometry (0.5 ug/test), Western Blot (1:10-1:100)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513582 309.00
Check your price
  • HLA-E Antibody (Monoclonal, MEM-E/06)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA119356
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Paraffin) (10 µg/ml), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119356 249.00
Check your price
  • HLA-E Antibody (Monoclonal, MEM-E/07)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119360
  • Size 100 µg
Applications
Flow Cytometry (Assay Dependent), Immunohistochemistry (Paraffin) (10 ug/ml), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119360 309.00
Check your price
  • HLA-E Antibody (Monoclonal, MEM-E/08)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA119362
  • Size 100 µg
Applications
Flow Cytometry (4 ug/ml), Immunoprecipitation (Assay Dependent)
Alternate names:
MHC class I antigen A*1; MHC class I antigen A*3
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119362 249.00
Check your price