1 - 30 of 30 products displayed
  • CCL3 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHC6034
  • Size 500 µg
Applications
ELISA (Assay Dependent)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHC6034 303.00
Check your price
  • CCL3 Polyclonal Antibody for Western Blot, IHC (P), ELISA and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547000
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (2-8 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547000 375.00
Check your price
  • CCL3 Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA523564
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523564 249.00
Check your price
  • CCL3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA138160
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA138160 425.00
Check your price
  • MIP-3 alpha Monoclonal Antibody for Western Blot
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516253
  • Size 100 ug
Applications
Western Blot (0.1-2 µg/ml)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1; CC chemokine ST38
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516253 349.00
Check your price
  • MIP-3 alpha Monoclonal Antibody for Western Blot, Flow, ELISA and Neu
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA523843
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Flow Cytometry (2.5 µg/10^6 cells), Neutralization (1-5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523843 275.00
Check your price
  • MIP-3 alpha Polyclonal Antibody for Western Blot, IF, ICC, IHC (P) and Neu
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547517
  • Size 100 µg
Applications
Immunocytochemistry (5-15 µg/ml), Immunofluorescence (5-15 µg/ml), Immunohistochemistry (Paraffin) (5-15 µg/ml), Neutralization (0.2-1.0 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547517 349.00
Check your price
  • MIP-3 alpha Monoclonal Antibody, FITC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate FITC
  • Catalog # MA523684
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523684 359.00
Check your price
  • MIP-3 alpha Monoclonal Antibody, PE conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate PE
  • Catalog # MA523583
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523583 279.00
Check your price
  • MIP-3 alpha Monoclonal Antibody, APC conjugate for Flow
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523563
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523563 249.00
Check your price
  • CCL3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA129045
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129045 375.00
Check your price
  • CCL3 Polyclonal Antibody for Western Blot, ELISA and Neu
  • Host Goat
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA127987
  • Size 100 µg
Applications
ELISA (1 µg/ml), Neutralization (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1; TY-5; SIS-alpha; L2G25B; Heparin-binding chemotaxis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127987 309.00
Check your price
  • MIP1 alpha Polyclonal Antibody for Western Blot, ELISA
  • Host Rabbit
  • Target Species Rat
  • Conjugate
  • Catalog # PA132967
  • Size 50µg
Applications
ELISA (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Small-inducible cytokine A3; Macrophage inflammatory protein 1-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132967 650.00
Check your price
  • CCL3 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA184295
  • Size 100 µg
Applications
ELISA (0.5-2 ug/ml), Western Blot (0.1-0.2ug/ml)
Alternate names:
TY-5; Small-inducible cytokine A3; SIS-alpha; Macrophage inflammatory protein 1-alpha; L2G25B; Heparin-binding chemotaxis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184295 475.00
Check your price
  • CCL3 Polyclonal Antibody for Western Blot and ELISA
  • Host Rabbit
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA128899
  • Size 50 µg
Applications
ELISA (0.5 µg/ml), Western Blot (0.1-0.2 µg/ml)
Alternate names:
TY-5; Small-inducible cytokine A3; SIS-alpha; Macrophage inflammatory protein 1-alpha; L2G25B; Heparin-binding chemotaxis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128899 375.00
Check your price
  • CCL3 Polyclonal Antibody for Western Blot, ELISA, Neu and FN
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA132966
  • Size 50 µg
Applications
ELISA (Assay-dependent), Functional Assay (Assay-dependent), Neutralization (Assay-dependent), Western Blot (Assay dependent)
Alternate names:
Small-inducible cytokine A3; Macrophage inflammatory protein 1-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132966 600.00
Check your price
  • CCL3 Polyclonal Antibody for IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532496
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532496 349.00
Check your price
  • MIP-3 alpha Monoclonal Antibody for Western Blot
  • Host Rat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516252
  • Size 100 ug
Applications
Western Blot (0.1-2 µg/ml)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine LARC; Beta-chemokine exodus-1; CC chemokine ST38
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516252 349.00
Check your price
  • CCL3 Monoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701097
  • Size 100 µg
Applications
Western Blot (0.1-1 ug/ml)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
701097 309.00
Check your price
  • CCL3 Polyclonal Antibody for ELISA
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA175246
  • Size 500 µg
Applications
ELISA (Assay Dependent)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA175246 1,500.00
Check your price
  • CCL3 Oligoclonal Antibody for Western Blot and IHC (P)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710127
  • Size 100 µg
Applications
Western Blot (2-5 ug/ml)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
710127 309.00
Check your price
  • MIP-3 alpha Monoclonal Antibody, FITC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate FITC
  • Catalog # MA523548
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine ST38; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523548 239.00
Check your price
  • MIP-3 alpha Monoclonal Antibody for Western Blot and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523856
  • Size 100 µg
Applications
Neutralization (1-5 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine ST38; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523856 275.00
Check your price
  • MIP-3 alpha Monoclonal Antibody for ELISA and Neu
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA523857
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 2-8 µg/ml (Capture)), Neutralization (3-15 µg/ml)
Alternate names:
Small-inducible cytokine A20; Macrophage inflammatory protein 3 alpha; Liver and activation-regulated chemokine; CC chemokine ST38; CC chemokine LARC; Beta-chemokine exodus-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523857 275.00
Check your price
  • CCL4 Monoclonal Antibody for ELISA
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # AHC6114
  • Size 500 µg
Applications
ELISA (Assay Dependent)
Alternate names:
T-cell activation protein 2; Small-inducible cytokine A4; SIS-gamma; Protein H400; PAT 744; Macrophage inflammatory protein 1-beta; MIP-1-beta(1-69); Lymphocyte activation gene 1 protein; HC21; G-26 T-lymphocyte-secreted protein; Small-inducible cytokine A4-like; Monocyte adherence-induced protein 5-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHC6114 307.00
Check your price
  • MIP-1 alpha for Ctrl
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP10923
  • Size 20 µg
Applications
Control (Assay Dependent)
Alternate names:
MIP-1 alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP10923 309.00
Check your price
  • CCL3 (human) for Ctrl
  • Host E. coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP8654
  • Size 20 µg
Applications
Control (Assay Dependent)
Alternate names:
Tonsillar lymphocyte LD78 alpha protein; Small-inducible cytokine A3; SIS-beta; PAT 464.1; Macrophage inflammatory protein 1-alpha; G0/G1 switch regulatory protein 19-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP8654 269.00
Check your price
  • CCL3 Monoclonal Antibody for Western Blot, IF and ICC
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA524364
  • Size 100 µg
Applications
Immunocytochemistry (8-25 µg/ml), Immunofluorescence (8-25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
TY-5; Small-inducible cytokine A3; SIS-alpha; Macrophage inflammatory protein 1-alpha; L2G25B; Heparin-binding chemotaxis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524364 275.00
Check your price
  • CCL3 Monoclonal Antibody, PE conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate PE
  • Catalog # MA523598
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
TY-5; Small-inducible cytokine A3; SIS-alpha; Macrophage inflammatory protein 1-alpha; L2G25B; Heparin-binding chemotaxis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523598 289.00
Check your price
  • CCL3 Monoclonal Antibody, APC conjugate for Flow
  • Host Rat
  • Target Species Mouse
  • Conjugate APC
  • Catalog # MA523647
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
TY-5; Small-inducible cytokine A3; SIS-alpha; Macrophage inflammatory protein 1-alpha; L2G25B; Heparin-binding chemotaxis protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523647 339.00
Check your price