1 - 2 of 2 products displayed
  • MECR Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534490
  • Size 100 ug
Applications
Western Blot (1-2 ug/mL)
Alternate names:
Nuclear receptor-binding factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534490 349.00
Check your price
  • MECR Polyclonal Antibody for Western Blot
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535243
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Nuclear receptor-binding factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535243 349.00
Check your price