1 - 30 of 30 products displayed
  • p38 MAPK alpha Antibody (Monoclonal)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Pig, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # 82382
  • Size 100 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:25), Immunofluorescence (1:25), Western Blot (1:1000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
82382 349.00
Check your price
  • ERK5 Antibody
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127890
  • Size 50 µg
Applications
Western Blot (1:2000)
Alternate names:
Extracellular signal-regulated kinase 5; Big MAP kinase 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127890 425.00
Check your price
  • p38 MAPK gamma Antibody (Monoclonal, 10E1)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA1100
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1:500)
Alternate names:
Stress-activated protein kinase 3; Mitogen-activated protein kinase p38 gamma; Extracellular signal-regulated kinase 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1100 349.00
Check your price
  • TAB1 Antibody (Monoclonal, 2D3)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0196
  • Size 100 µL
Applications
ELISA (Assay Dependent), Western Blot (1:5000-1:10,000)
Alternate names:
TGF-beta-activated kinase 1-binding protein 1; Mitogen-activated protein kinase kinase kinase 7-interacting protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0196 309.00
Check your price
  • ERK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA137640
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137640 375.00
Check your price
  • ERK2 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532396
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532396 275.00
Check your price
  • ERK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA129167
  • Size 500 µL
Applications
Western Blot (1:1000)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA129167 349.00
Check your price
  • ERK2 Antibody (Monoclonal, 6F8)
  • Host Mouse
  • Target Species Dog, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1099
  • Size 100 µg
Applications
ChIP assay (1-3 ul), Flow Cytometry (1-10 µg/ml), Immunocytochemistry (1:200), Immunofluorescence (1:200), Immunohistochemistry (Paraffin) (1:100-1:500), Immunoprecipitation (2 µg/500 µl), Western Blot (1:1000)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1099 349.00
Check your price
  • JNK1 Antibody (Oligoclonal, 2HCLC)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 710075
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (0.5–3 ug/ml)
Alternate names:
c-Jun N-terminal kinase 1; Stress-activated protein kinase JNK1; Stress-activated protein kinase 1c; JNK-46
Catalog # Size Price (AUD) Your Price (AUD) Qty
710075 309.00
Check your price
  • ERK1/ERK2 Antibody (Monoclonal, 3A1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0178
  • Size 100 µL
Applications
ELISA (Assay Dependent), Western Blot (0.1 µg/ml)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0178 309.00
Check your price
  • p38 MAPK alpha Antibody (Monoclonal, p38-3F11)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # 331300
  • Size 100 µg
Applications
ELISA (0.1-1.0 ug/ml), Immunocytochemistry (5-20 ug/ml), Immunofluorescence (5-20 ug/ml), Western Blot (1.0 ug/ml)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
331300 305.00
Check your price
  • ERK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA121016
  • Size 500 µL
Applications
ChIP assay (Assay-Dependent), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (Assay dependent)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121016 349.00
Check your price
  • JNK1 Antibody (Monoclonal, 2H15L8)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # 701048
  • Size 100 µg
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Western Blot (1-3 ug/ml)
Alternate names:
c-Jun N-terminal kinase 1; Stress-activated protein kinase JNK1; Stress-activated protein kinase 1c; JNK-46
Catalog # Size Price (AUD) Your Price (AUD) Qty
701048 309.00
Check your price
  • MKP4 Antibody (Monoclonal, AF11B5)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517233
  • Size 100 µL
Applications
ELISA (Assay Dependent), Western Blot (1:2000)
Alternate names:
Mitogen-activated protein kinase phosphatase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517233 349.00
Check your price
  • ERK1/ERK2 Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44654G
  • Size 100 µL
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Immunohistochemistry (Paraffin) (1:10-1:100), Western Blot (1:500-1:2000)
Alternate names:
p44-ERK1; Microtubule-associated protein 2 kinase; MNK1; MAP kinase isoform p44; Insulin-stimulated MAP2 kinase; Extracellular signal-regulated kinase 1; ERT2; Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
44654G 304.00
Check your price
  • p38 MAPK alpha Antibody (Monoclonal, 6A1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0126
  • Size 100 µL
Applications
Western Blot (4 µg/ml)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0126 309.00
Check your price
  • p38 MAPK alpha Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534769
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:500-1:3000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534769 349.00
Check your price
  • JNK2 Antibody (Monoclonal, 341P16)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA121411
  • Size 50 µg
Applications
ELISA (Assay dependent), Western Blot (1 µg/ml)
Alternate names:
c-Jun N-terminal kinase 2; Stress-activated protein kinase JNK2; Stress-activated protein kinase 1a; JNK-55; p54-alpha; SAPK-alpha
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121411 399.00
Check your price
  • JNK1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44690G
  • Size 100 µL
Applications
Flow Cytometry (3-5 µg/million cells), Immunohistochemistry (Paraffin) (1:20-1:200), Western Blot (1-2 µg/ml)
Alternate names:
p54 gamma; c-Jun N-terminal kinase 1; Stress-activated protein kinase JNK1; SAPK gamma; Stress-activated protein kinase 1c; JNK-46
Catalog # Size Price (AUD) Your Price (AUD) Qty
44690G 309.00
Check your price
  • JIP1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # 345200
  • Size 100 µg
Applications
Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Mitogen-activated protein kinase 8-interacting protein 1; JNK MAP kinase scaffold protein 1; Islet-brain 1; Islet-brain-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
345200 309.00
Check your price
  • ERK2 Antibody (Monoclonal, 1A1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0193
  • Size 100 µL
Applications
Immunoprecipitation (2 µl ), Western Blot (1:2000)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0193 309.00
Check your price
  • ERK2 Antibody (Monoclonal, 1A8)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # LFMA0179
  • Size 100 µL
Applications
ELISA (Assay Dependent), Western Blot (1:1000)
Alternate names:
Mitogen-activated protein kinase 2; MAP kinase isoform p42; Extracellular signal-regulated kinase 2; ERT1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0179 309.00
Check your price
  • JNK1 Antibody (Monoclonal, 4A2G12)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # 359800
  • Size 100 µg
Applications
Flow Cytometry (3-5 ug x 10^6 cells), Immunocytochemistry (1-3 ug/ml), Immunofluorescence (1-3 ug/ml), Western Blot (1-3 ug/ml)
Alternate names:
c-Jun N-terminal kinase 1; Stress-activated protein kinase JNK1; Stress-activated protein kinase 1c; JNK-46
Catalog # Size Price (AUD) Your Price (AUD) Qty
359800 309.00
Check your price
  • MKP2 Antibody (Monoclonal, AFPK7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA517225
  • Size 100 µL
Applications
ELISA (Assay Dependent), Western Blot (1:2000)
Alternate names:
Mitogen-activated protein kinase phosphatase 2; Dual specificity protein phosphatase hVH2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517225 349.00
Check your price
  • MEK3 Antibody (Monoclonal, 3G2)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA517216
  • Size 100 µL
Applications
Western Blot (2 ug/ml)
Alternate names:
MAPK/ERK kinase 3; Stress-activated protein kinase kinase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517216 349.00
Check your price
  • DUSP10 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA131512
  • Size 50 µg
Applications
Western Blot (0.5-2 µg/ml)
Alternate names:
Mitogen-activated protein kinase phosphatase 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131512 399.00
Check your price
  • DUSP16 Antibody
  • Host Rabbit
  • Target Species Bovine, Human, Mouse, Non-human primate
  • Conjugate Unconjugated
  • Catalog # PA523140
  • Size 100 ug
Applications
ChIP assay (1-3 ul), Immunohistochemistry (Paraffin) (10 µg/ml), Western Blot (1-3 µg/ml)
Alternate names:
Mitogen-activated protein kinase phosphatase 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523140 349.00
Check your price
  • MKP4 Antibody
  • Host Chicken
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA127644
  • Size 50 µg
Applications
Western Blot (1:2000)
Alternate names:
Mitogen-activated protein kinase phosphatase 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA127644 425.00
Check your price
  • p38 MAPK alpha Antibody (Monoclonal, G.876.7)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515204
  • Size 100 µL
Applications
Western Blot (1:2000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515204 275.00
Check your price
  • p38 MAPK alpha Antibody (Monoclonal, J.491.1)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Pig, Rat, Yeast
  • Conjugate Unconjugated
  • Catalog # MA515116
  • Size 100 µL
Applications
Flow Cytometry (1:200), Immunocytochemistry (1:25), Immunofluorescence (1:25), Western Blot (1:1000)
Alternate names:
Stress-activated protein kinase 2a; Mitogen-activated protein kinase p38 alpha; MAX-interacting protein 2; MAP kinase MXI2; Cytokine suppressive anti-inflammatory drug-binding protein; CRK1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515116 275.00
Check your price