1 - 2 of 2 products displayed
  • Phospho-Tau (Ser202, Thr205) Antibody, Biotin (Monoclonal, AT8)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin See Additional Formats Unconjugated
  • Catalog # MN1020B
  • Size 50 ug
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MN1020B 499.00
Check your price
  • Phospho-Tau (Ser202, Thr205) Antibody (Monoclonal, AT8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated See Additional Formats Biotin
  • Catalog # MN1020
  • Size 100 ug
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MN1020 499.00
Check your price